Skip to Content
Merck
All Photos(2)

Key Documents

W267020

Sigma-Aldrich

p-Anisaldehyde

natural, FG

Synonym(s):

4-Methoxybenzaldehyde, Aubépine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2670
Beilstein:
471382
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.015
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

4.7 (vs air)

form

liquid

refractive index

n20/D 1.573 (lit.)

bp

248 °C (lit.)

mp

−1 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise; cherry; creamy; floral; balsamic; sweet; vanilla

SMILES string

[H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Vanilla, fennel, star anise, cranberry, black currant, cinnamon, basil.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natcha Panthama et al.
Archives of pharmacal research, 38(5), 585-590 (2014-06-11)
Two new angular types of azaphilones, isochromophilonol (1) and ochrephilonol (2), together with ten known compounds (3-12), were isolated from Chaetomium cupreum RY202. Their structures were established on the basis of spectroscopic data and the absolute configurations of 1 and
Wen-Fei He et al.
Fitoterapia, 96, 109-114 (2014-04-29)
Two new renieramycin-type bistetrahydroisoquinolinequinone alkaloids, fennebricins A (1) and B (5), and one new isoquinolinequinone alkaloid, N-formyl-1,2-dihydrorenierol (7), were isolated from the skin of the South China Sea nudibranch Jorunna funebris and its possible sponge-prey Xestospongia sp., together with eight
Navjot Kaur et al.
Antonie van Leeuwenhoek, 106(2), 301-307 (2014-05-27)
A Gram-positive, yellow pigmented strain, BKS 3-46(T) was isolated from a soil sample collected from the rhizosphere of Ficus benghalensis (banyan tree) in Bhitarkanika mangrove forest, in the Indian state of Odisha, and subjected to polyphasic taxonomic study. The 16S
Zhuozhi Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(45), 14637-14640 (2014-10-07)
Controlled release of odorous molecules is the key to digital scent technology which will add another dimension to electronics. Photorelease is a cold mechanism that promises better temporal and spatial control than thermal release. Herein we report a novel material
Haruki Mizoguchi et al.
Organic & biomolecular chemistry, 13(21), 5955-5963 (2015-05-01)
Copper-catalyzed 6-endo cyclization of N-propargylic β-enaminocarbonyls was developed for the synthesis of oxidation-labile 1,6-dihydropyridines. This synthetic method allows flexible and regio-defined assembly of various substituents at the N1, C2, C3, C4, and C6 positions of 1,6-dihydropyridines under mild conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service