Skip to Content
Merck
All Photos(3)

Key Documents

W310212

Sigma-Aldrich

Isovaleric acid

greener alternative

natural, ≥98%, FG

Synonym(s):

3-Methylbutanoic acid, 3-Methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOH
CAS Number:
Molecular Weight:
102.13
FEMA Number:
3102
Beilstein:
1098522
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.008
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor pressure

0.38 mmHg ( 20 °C)

Assay

≥98%

autoignition temp.

824 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.403 (lit.)

bp

175-177 °C (lit.)

mp

−29 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cheese; fatty; acidic; sour

SMILES string

CC(C)CC(O)=O

InChI

1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

GWYFCOCPABKNJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - Pensky-Martens closed cup

Flash Point(C)

80 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jechan Lee et al.
Environmental pollution (Barking, Essex : 1987), 230, 81-86 (2017-06-27)
This study lays great emphasis on establishing a reliable analytical platform to quantify and specify volatile fatty acids (VFAs) in the aqueous phase by derivatizing VFAs into their corresponding alkyl esters via thermally-induced rapid esterification (only 10 s reaction time). To
Georg F Hoffmann et al.
European journal of pediatrics, 163(2), 76-80 (2004-01-10)
The lack of epidemiological data on the frequency and/or burden of organic acidurias (OA) and mitochondrial fatty acid transport and oxidation disorders (mtFATOD) is one reason for hesitation to expand newborn screening (NBS) by tandem mass spectrometry (MS-MS). From 1999
Eilhann E Kwon et al.
Environmental research, 183, 109199-109199 (2020-02-07)
In this study, it is introduced a sustainable synthetic route of alkyl esters, considered value-added industrial chemicals and fuels, from volatile fatty acids (VFAs) that can potentially be generated from organic waste. In the presence of a porous carbon material
M F Silva et al.
Clinical biochemistry, 34(8), 635-638 (2002-02-19)
The effect of administration of the antiepileptic drug valproate (VPA), on the composition of the plasma acylcarnitine profile (including free carnitine) was investigated. Plasma samples were obtained from 18 individuals (13 males:5 females; 15-65 y) on long-term treatment with VPA
Wolfgang R Mukabana et al.
Journal of chemical ecology, 38(3), 235-244 (2012-03-20)
Estimating the biting fraction of mosquitoes is of critical importance for risk assessment of malaria transmission. Here, we present a novel odor-based tool that has been rigorously assessed in semi-field assays and traditional African villages for estimating the number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service