Skip to Content
Merck
All Photos(1)

Key Documents

W286612

Sigma-Aldrich

Phenethyl phenylacetate

natural, ≥98%, FG

Synonym(s):

Benzyl carbinyl phenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O2
CAS Number:
Molecular Weight:
240.30
FEMA Number:
2866
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.707
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

honey; floral; rose; sweet

SMILES string

O(CCc2ccccc2)C(=O)Cc1ccccc1

InChI

1S/C16H16O2/c17-16(13-15-9-5-2-6-10-15)18-12-11-14-7-3-1-4-8-14/h1-10H,11-13H2

InChI key

ZOZIRNMDEZKZHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service