Skip to Content
Merck
All Photos(1)

Key Documents

W267112

Sigma-Aldrich

2-Methoxy-4-methylphenol

natural, 97%, FG

Synonym(s):

2-Hydroxy-5-methylanisole, 2-Methoxy-p-cresol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, Creosol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)OH
CAS Number:
Molecular Weight:
138.16
FEMA Number:
2671
Beilstein:
1862340
EC Number:
Council of Europe no.:
175c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.007
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97%

refractive index

n20/D 1.537 (lit.)

bp

221-222 °C (lit.)

mp

5 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

clove; leathery; smoky; spicy; smoky; sweet; vanilla

SMILES string

COc1cc(C)ccc1O

InChI

1S/C8H10O2/c1-6-3-4-7(9)8(5-6)10-2/h3-5,9H,1-2H3

InChI key

PETRWTHZSKVLRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methoxy-4-methylphenol is a phenolic flavor compound reported in commercial liquid smoke flavoring and wood smoke.

Biochem/physiol Actions

Taste at 1.0-1.25 ppm

Other Notes

Natural occurrence: Coffee, gruyere cheese, pork, beer, rum, bourbon whiskey, malt, sherry, cocoa, tea, mushroom, bourbon vanilla, green mate, jasmine flowers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of a commercial liquid smoke flavoring by means of gas chromatography/mass spectrometry and Fourier transform infrared spectroscopy.
Guillen MD, et al.
Journal of Agricultural and Food Chemistry, 43(2), 463-468 (1995)
Day-Shin Hsu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(10), 3121-3131 (2010-02-02)
An efficient and short entry to polyfunctionalized linear triquinanes from 2-methoxyphenols is described by utilizing the following chemistry. The Diels-Alder reactions of masked o-benzoquinones, derived from 2-methoxyphenols, with cyclopentadiene afford tricyclo[5.2.2.0(2,6)]undeca-4,10-dien-8-ones. Photochemical oxa-di-pi-methane (ODPM) rearrangements and 1,3-acyl shifts of the
Anthea L Fudge et al.
Journal of agricultural and food chemistry, 60(1), 52-59 (2011-12-02)
In this study, the suitability of mid-infrared (MIR) spectroscopy, combined with principal component analysis (PCA) and linear discriminant analysis (LDA), was evaluated as a rapid analytical technique to identify smoke tainted wines. Control (i.e., unsmoked) and smoke-affected wines (260 in
The flavor chemistry of wood smoke.
Maga JA
Food Reviews International, 3(1-2), 139-183 (1987)
N Ogata et al.
Pharmacology, 51(3), 195-204 (1995-09-01)
Wood creosote, principally a mixture of non-, alkyl- and/or alkoxy-substituted phenolic compounds, was orally administered to adult male volunteers to determine its metabolites and pharmacokinetic parameters. After a 133-mg single dose, its major constituents (i.e. phenol 15 mg, guaiacol 32

Articles

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service