Skip to Content
Merck
All Photos(1)

Key Documents

W259306

Sigma-Aldrich

Indole

≥99%, FG

Synonym(s):

1H-Benzo[b]pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
FEMA Number:
2593
Beilstein:
107693
EC Number:
Council of Europe no.:
560
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.007
NACRES:
NA.21
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥99%

impurities

≤250 ppm Hexane, residual (solvent)

bp

253-254 °C (lit.)

mp

51-54 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

pungent; floral; animal

SMILES string

c1ccc2[nH]ccc2c1

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indole is one of the main flavor constituents of green tea.[1]

Biochem/physiol Actions

Taste at 0.3-2 ppm

Other Notes

Natural occurrence: Burley tobacco, butter, coffee, dried bonito, egg, fish, malt, rum, tea, and wine.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Further Investigation of Flavor Constituents in Manufactured Green Tea
Yamanishi T, et al.
Agricultural and Biological Chemistry, 34(4), 599-608 (1970)
Hélène Gérard et al.
The Journal of organic chemistry, 78(18), 9233-9242 (2013-08-07)
The chemo-, regio-, and stereoselectivities of multicomponent [4 + 2]/[3 + 2] domino cycloaddition reactions involving nitroindole derivatives with vinylethers and acrylates are studied computationnally and compared to experimental results. In this process, the nitroarene first reacts as an electron-deficient
Xueqing Geng et al.
The Plant cell, 24(11), 4763-4774 (2012-12-04)
The phytotoxin coronatine (COR) promotes various aspects of Pseudomonas syringae virulence, including invasion through stomata, growth in the apoplast, and induction of disease symptoms. COR is a structural mimic of active jasmonic acid (JA) conjugates. Known activities of COR are
Total synthesis of the tremorgenic indole diterpene paspalinine.
Masaru Enomoto et al.
Angewandte Chemie (International ed. in English), 51(51), 12833-12836 (2012-11-09)
Facile access to 3-acylindoles through palladium-catalyzed addition of indoles to nitriles: the one-pot synthesis of indenoindolones.
Yuanhong Ma et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(4), 1189-1193 (2012-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service