Skip to Content
Merck
All Photos(2)

Key Documents

I5605

Sigma-Aldrich

Indoline

ReagentPlus®, 99%

Synonym(s):

1-Azaindan, 2,3-Dihydroindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N
CAS Number:
Molecular Weight:
119.16
Beilstein:
111915
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.592 (lit.)

bp

220-221 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2N1

InChI

1S/C8H9N/c1-2-4-8-7(3-1)5-6-9-8/h1-4,9H,5-6H2

InChI key

LPAGFVYQRIESJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Inhibitors of NOD1-Induced Nuclear Factor-κB Activation
  • Sphingosine-1-phosphate 4(S1P4) receptor antagonists
  • Cytotoxic cell cycle inhibitors
  • 2-Aminopyridines
  • PET agent for imaging of protein kinase C (PKC)
  • Sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
  • α4β2-Nicotinic acetylcholine receptor-selective partial agonists
  • mGlu4 positive allosteric modulators
  • Bacterial biofilm inhibitors
  • Serotonin 5-HT6 receptor antagonists

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matej Baláž et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-22)
Performing solution-phase oximation reactions with hydroxylamine hydrochloride (NH2OH·HCl) carries significant risk, especially in aqueous solutions. In the present study, four N-substituted indole-3-carboxaldehyde oximes were prepared from the corresponding aldehydes by solvent-free reaction with NH2OH·HCl and a base (NaOH or Na2CO3)
Toshiharu Noji et al.
Organic letters, 15(8), 1946-1949 (2013-04-04)
A benzyne-mediated synthesis of substituted indolines and carbazoles was developed. The reaction includes generation of benzyne using Mg(TMP)2·2LiCl as a base, cyclization, and trapping the resulting organomagnesium intermediate with an electrophile to provide a series of substituted indolines and carbazoles
Manas K Ghorai et al.
The Journal of organic chemistry, 78(8), 3867-3878 (2013-04-04)
A practical approach for the synthesis of 3-substituted indolines via regio- and stereoselective SN2-type ring-opening of 2-(2-halophenyl)-N-tosylaziridines with heteroatomic nucleophiles (O, N, and S) followed by palladium-catalyzed intramolecular C-N cyclization is reported in excellent yields (up to >99%) and enantiomeric
Tanguy Saget et al.
Organic letters, 15(6), 1354-1357 (2013-03-05)
The synthesis of cyclopropyl spiroindolines is described using an intramolecular palladium(0)-catalyzed C-H functionalization of a methine C(sp(3))-H bond. This transformation can be coupled with intermolecular Suzuki couplings or direct arylations of heteroaromatics to access functionalized indoline scaffolds in a single
A diversity-oriented approach to spiroindolines: post-Ugi gold-catalyzed diastereoselective domino cyclization.
Sachin G Modha et al.
Angewandte Chemie (International ed. in English), 51(38), 9572-9575 (2012-08-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service