Skip to Content
Merck
All Photos(1)

Documents

W245801

Sigma-Aldrich

Ethyl salicylate

≥99%, FCC, FG

Synonym(s):

Ethyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
FEMA Number:
2458
Beilstein:
907659
EC Number:
Council of Europe no.:
432
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39024568
PubChem Substance ID:
Flavis number:
9.748
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

0.05 mmHg ( 25 °C)

Assay

≥99%

refractive index

n20/D 1.522 (lit.)

bp

234 °C (lit.)

mp

1 °C (lit.)

density

1.131 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy; balsamic; minty; sweet; wintergreen

SMILES string

CCOC(=O)c1ccccc1O

InChI

1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3

InChI key

GYCKQBWUSACYIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Collins et al.
Annals of the rheumatic diseases, 43(3), 411-415 (1984-06-01)
A topically applied rubifacient delivered by aerosol (Deep-Heat) was studied. After spray application to the forearms of volunteers, without massage, the erythema produced was measured by thermography and correlated with the concentration of 2 salicylate components of the mixture found
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S397-S401 (2007-11-21)
A toxicological and dermatological review of ethyl salicylate when used as a fragrance ingredient is presented.
Amnon C Sintov et al.
Journal of controlled release : official journal of the Controlled Release Society, 79(1-3), 113-122 (2002-02-21)
The aim of this study was to investigate the percutaneous penetration and dermal metabolism of a new potential anti-acne prodrug--TU-2100 [bis(o-carboxyphenyl ethyl ester)nonanedioate] in guinea pigs. The fluxes of this agent through excised skin after applications of TU-2100 gels at
Bryan J McCullough et al.
Rapid communications in mass spectrometry : RCM, 25(10), 1445-1451 (2011-04-20)
The design and development of a novel extractive electrospray ionisation (EESI) device for on-line reaction monitoring is described. The EESI apparatus uses a secondary, grounded nebuliser to produce an analyte aerosol and a Venturi pump is then used to transfer
Mauricio Filippa et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 969-973 (2008-07-25)
The complexation of methyl salicylate (MS) and ethyl salicylate (ES), non-steroidal analgesic, anti-inflammatory and antirrheumatic drugs with beta-cyclodextrin (betaCD) has been studied from thermodynamic and structural points of view. The complexation with betaCD has been investigated using reversed-phase liquid chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service