Skip to Content
Merck
All Photos(1)

Key Documents

P11255

Sigma-Aldrich

PTCDA

97%

Synonym(s):

Perylene-3,4,9,10-tetracarboxylic dianhydride, Pigment Red 224

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H8O6
CAS Number:
Molecular Weight:
392.32
Beilstein:
57831
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

Band gap: 2.1 eV

Assay

97%

form

powder

mp

>300 °C

Orbital energy

HOMO -6.8 eV 
LUMO -4.7 eV 

OPV Device Performance

ITO/CuPc/PTCDA/In

  • Short-circuit current density (Jsc): 2 mA/cm2
  • Open-circuit voltage (Voc): 0.55 V
  • Fill Factor (FF): 0.35
  • Power Conversion Efficiency (PCE): 1.8 %

semiconductor properties

N-type (mobility=10−4 cm2/V·s)

SMILES string

O1C(=O)c2c3c4c(c5c6c(c4cc2)ccc7c6c(cc5)C(=O)OC7=O)ccc3C1=O

InChI

1S/C24H8O6/c25-21-13-5-1-9-10-2-6-15-20-16(24(28)30-23(15)27)8-4-12(18(10)20)11-3-7-14(22(26)29-21)19(13)17(9)11/h1-8H

InChI key

CLYVDMAATCIVBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perylene-3,4,9,10-tetracarboxylic dianhydride (PTCDA) is a perylene derivative which forms highly crystalline films and dyes that can be used for a majority of electronic and opto-electronic applications. It provides a high electron mobility due to its low intermolecular distance which results in π-π conjugation.

Application

Polycondensation of PTCDA with polypropylene glycol (PPG) based diamine can be utilized for the development of perlyene diimides which have a potential use as n-type semiconductors in organic photovoltaics. PTCDA can be used as a light absorbing monomer for the fabrication of bichromophobic light harvesting antenna systems. It can also be used to prepare 3, 4, 9, 10-perylene tetracarboxylic acid-aromatic fluorophores dye for a highly reversible fluorescence switching on different substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Richard Prabakar et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(24), 1902129-1902129 (2020-01-01)
Herein, graphite is proposed as a reliable Ca2+-intercalation anode in tetraglyme (G4). When charged (reduced), graphite accommodates solvated Ca2+-ions (Ca-G4) and delivers a reversible capacity of 62 mAh g-1 that signifies the formation of a ternary intercalation compound, Ca-G4·C72. Mass/volume
Electrochemical and optical studies of 1,4-diaminoanthraquinone for solar cell applications
Brovelli, F.; Rivas, B.; Bernede, J.; et al.
Polymer Bull., 58, 521-527 (2007)
Tunable and highly efficient light-harvesting antenna systems based on 1, 7-perylene-3, 4, 9, 10-tetracarboxylic acid derivatives.
Dubey RK, et al.
Chemical Science, 7(6), 3517-3532 (2016)
Sylvie Rangan et al.
The journal of physical chemistry. B, 122(2), 534-542 (2017-06-21)
The adsorption geometry and energy alignment at the PTCDA/TiO
Fanfan Du et al.
Cellular and molecular bioengineering, 10(5), 357-370 (2017-10-11)
Intracellular delivery is a key step for many applications in medicine and for investigations into cellular function. This is particularly true for immunotherapy, which often requires controlled delivery of antigen and adjuvants to the cytoplasm of immune cells. Due to

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service