Skip to Content
Merck
All Photos(2)

Key Documents

932353

Sigma-Aldrich

PTCDI

Synonym(s):

3,4,9,10-Perylenetetracarboxylic acid diimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H10N2O4
CAS Number:
Molecular Weight:
390.35
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
sublimed grade

grade

sublimed grade

Quality Level

loss

0.5% TGA, > 450 °C (weight loss)

SMILES string

[nH]1[c](c2c3c4c(c5c6c7c([c]([nH][c](c7cc5)=O)=O)ccc6c4cc2)ccc3[c]1=O)=O

InChI

1S/C24H10N2O4/c27-21-13-5-1-9-10-2-6-15-20-16(24(30)26-23(15)29)8-4-12(18(10)20)11-3-7-14(22(28)25-21)19(13)17(9)11/h1-8H,(H,25,27,28)(H,26,29,30)

Application

This TADF emitter is a sublimed high purity material, solution processable 3,4,9,10-perylenetetracarboxylic diimide has been reported for use in:
  • photovoltaics
  • thermoelectricity (induced by the Soret effect )
  • batteries for energy storage
  • photocatalysis for water purification or selective solar fuel production from CO2
  • chemosensing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Wang et al.
ACS nano, 14(5), 5998-6006 (2020-05-10)
The visual aesthetic that involves color, brightness, and glossiness is of great importance for building integrated photovoltaics. Semitransparent organic solar cells (ST-OSCs) are thus considered as the most promising candidate due to their superiority in transparency and efficiency. However, the
Strongly coupled perylene bisimide/reduced graphene oxide as organic cathode materials for lithium ion batteries
W.Deng et al.
Electrochimica Acta, 282, 24-29 (2018)
Di Liu et al.
Advanced materials (Deerfield Beach, Fla.), 28(33), 7284-7290 (2016-06-17)
A self-assembled perylene-3,4,9,10-tetracarboxylic diimide(PDINH) supramolecular system consisting of all-organic PDINH molecule building blocks through non-covalent interactions works as a visible light photocatalyst with high activity.
Fully Solution-Processed Small Molecule Semitransparent Solar Cells: Optimization of Transparent Cathode Architecture and Four Absorbing Layers
J.Min et al.
Advanced Functional Materials , 26, 4543-4550 (2016)
Qinglin Jiang et al.
Advanced materials (Deerfield Beach, Fla.), 32(45), e2002752-e2002752 (2020-09-15)
Low-cost, non-toxic, abundant organic thermoelectric materials are currently under investigation for use as potential alternatives for the production of electricity from waste heat. While organic conductors reach electrical conductivities as high as their inorganic counterparts, they suffer from an overall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service