Skip to Content
Merck
All Photos(1)

Key Documents

N4002

Sigma-Aldrich

2-Naphthaleneacetic acid

99%

Synonym(s):

2-Naphthylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CO2H
CAS Number:
Molecular Weight:
186.21
Beilstein:
973396
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

141-143 °C (lit.)

SMILES string

OC(=O)Cc1ccc2ccccc2c1

InChI

1S/C12H10O2/c13-12(14)8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7H,8H2,(H,13,14)

InChI key

VIBOGIYPPWLDTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Darío Falcone et al.
Langmuir : the ACS journal of surfaces and colloids, 20(14), 5732-5737 (2006-02-08)
The kinetics of hydrolysis of 2-naphthyl acetate (2-NA) catalyzed by alpha-chymotrypsin (alpha-CT), in reverse micellar solutions formed by glycerol (GY)-water (38% v/v) mixture/sodium bis(2-ethylhexyl)sulfosuccinate (AOT)/n-heptane has been determined by spectroscopic measurements. To compare the efficiency of this reaction with that
T S Emudianughe et al.
Xenobiotica; the fate of foreign compounds in biological systems, 17(7), 823-828 (1987-07-01)
1. The influence of dose size upon the metabolic conjugation of 2-naphthylacetic acid with various amino acids and glucuronic acid has been studied in the guinea pig, mouse and hamster. 2. Guinea pigs conjugated 2-naphthylacetic acid with glycine and glucuronic
Misaki Nakai et al.
Inorganic chemistry, 45(7), 3048-3056 (2006-03-28)
Two new dinuclear Ru(III) complexes containing naphthalene moieties, K[Ru2(dhpta)(mu-O2CCH2-1-naph)2] (1) and K[Ru2(dhpta)(mu-O2CCH2-2-naph)2] (2) (H5dhpta = 1,3-diamino-2-hydroxypropane-N,N,N',N'-tetraacetic acid, naph-1-CH2CO2H = 1-naphthylacetic acid, naph-2-CH2CO2H = 2-naphthylacetic acid), were synthesized. Complex 2 crystallized as an orthorhombic system having a space group of Pbca
Hiroshi Ikeda et al.
Organic letters, 5(10), 1625-1627 (2003-05-09)
[reaction: see text] 6(A),6(D)-Bispyridinio-appended gamma-cyclodextrin effectively enhanced the excimer fluorescence of 2-naphthylacetate. This increase derived from the increase of formation of the 1:2 complex between the cation-charged gamma-cyclodextrin and 2-naphthylacetate by the electrostatic interaction between the host and the guest.
E Abuin et al.
Journal of colloid and interface science, 283(1), 87-93 (2005-02-08)
A study has been made of the effect of urea upon the hydrolysis of 2-naphthyl acetate (2-NA) catalyzed by lipase from Rhizopus arrhizus in AOT-heptane-water reverse micellar solutions at pH 7. The partition constants, K, of 2-NA between n-heptane and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service