Skip to Content
Merck
All Photos(2)

Key Documents

317918

Sigma-Aldrich

1-Naphthaleneacetic acid

technical grade

Synonym(s):

α-Naphthaleneacetic acid, 1-Naphthylacetic acid, NAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O2
CAS Number:
Molecular Weight:
186.21
Beilstein:
1308415
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

≥90.0% (GC)

form

lumps
powder

SMILES string

OC(=O)Cc1cccc2ccccc12

InChI

1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)

InChI key

PRPINYUDVPFIRX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amalia Riga et al.
PLoS genetics, 17(10), e1009856-e1009856 (2021-10-22)
The conserved adapter protein Scribble (Scrib) plays essential roles in a variety of cellular processes, including polarity establishment, proliferation, and directed cell migration. While the mechanisms through which Scrib promotes epithelial polarity are beginning to be unraveled, its roles in
Ravi Das et al.
Science advances, 7(38), eabg4617-eabg4617 (2021-09-18)
[Figure: see text].
Michael A Q Martinez et al.
G3 (Bethesda, Md.), 10(1), 267-280 (2019-11-16)
As developmental biologists in the age of genome editing, we now have access to an ever-increasing array of tools to manipulate endogenous gene expression. The auxin-inducible degradation system allows for spatial and temporal control of protein degradation via a hormone-inducible
Zaigham Shahzad et al.
Nature communications, 11(1), 218-218 (2020-01-12)
Maintaining the right balance between plasticity and robustness in biological systems is important to allow adaptation while maintaining essential functions. Developmental plasticity of plant root systems has been the subject of intensive research, but the mechanisms underpinning robustness remain unclear.
Yu-Tong Jiang et al.
The New phytologist, 225(4), 1606-1617 (2019-10-01)
Two types of tonoplast proton pumps, H+ -pyrophosphatase (V-PPase) and the H+ -ATPase (V-ATPase), establish the proton gradient that powers molecular traffic across the tonoplast thereby facilitating turgor regulation and nutrient homeostasis. However, how proton pumps regulate development remains unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service