Skip to Content
Merck
All Photos(1)

Key Documents

M13807

Sigma-Aldrich

4-Methoxycinnamic acid, predominantly trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
178.18
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid crystal

mp

170-173 °C (lit.)

transition temp

crystalline phase to nematic phase 173.5 °C
nematic phase to isotropic phase 190 °C

SMILES string

COc1ccc(\C=C\C(O)=O)cc1

InChI

1S/C10H10O3/c1-13-9-5-2-8(3-6-9)4-7-10(11)12/h2-7H,1H3,(H,11,12)/b7-4+

InChI key

AFDXODALSZRGIH-QPJJXVBHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha Cook et al.
The Australasian journal of dermatology, 43(2), 133-135 (2002-05-02)
We report a case of photoallergic contact dermatitis to two sunscreen agents, methoxycinnamate and oxybenzone, occurring in a 6-year-old Asian boy.
H V Meyers et al.
Carbohydrate research, 197, 15-32 (1990-03-25)
This study completes the spectroscopic basis for a novel oligosaccharide microanalytical method, wherein a derivatization sequence provides monosaccharide subunits bearing two types of exciton-coupling chromophore groups ("bichromophoric") for circular dichroic spectroscopy, namely 4-bromobenzoate (lambda max 245 nm) and 4-methoxycinnamate (lambda
Sirintorn Yibchok-anun et al.
Basic & clinical pharmacology & toxicology, 102(5), 476-482 (2008-03-19)
This study investigated the effect of p-methoxycinnamic acid (p-MCA) on plasma glucose and insulin concentrations in normal and streptozotocin-induced diabetic rats. In both fasting and glucose-loading conditions, an oral administration of p-MCA (40-100 mg/kg) significantly decreased plasma glucose and also
S Adisakwattana et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 43(11), 766-773 (2011-10-20)
p-Methoxycinnamic acid (p-MCA) is a cinnamic acid derivative that shows various pharmacologic actions such as hepatoprotective and antihyperglycemic activities. The present study was to elucidate the mechanisms by which p-MCA increases [Ca²⁺]i and insulin secretion in INS-1 cells. p-MCA (100
So Ra Kim et al.
British journal of pharmacology, 135(5), 1281-1291 (2002-03-06)
1. We previously reported that four new phenylpropanoid glycosides and six known cinnamate derivatives isolated from roots of Scrophularia buergeriana Miquel (Scrophulariaceae) protected cultured cortical neurons from neurotoxicity induced by glutamate. Here, we have investigated the structure-activity relationships in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service