Skip to Content
Merck
All Photos(1)

Key Documents

M10050

Sigma-Aldrich

3-Methoxybenzamide

97%

Synonym(s):

m-Anisamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CONH2
CAS Number:
Molecular Weight:
151.16
Beilstein:
2206857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

132.5-135.5 °C (lit.)

SMILES string

COc1cccc(c1)C(N)=O

InChI

1S/C8H9NO2/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3,(H2,9,10)

InChI key

VKPLPDIMEREJJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Le Page et al.
Advances in experimental medicine and biology, 419, 203-207 (1997-01-01)
Nitric oxide (NO) is a potent mediator involved in many biological functions including macrophage cytotoxicity and non-specific immunity against parasites, bacteria and viruses. Murine macrophages possess the capacity to express the inducible NO synthase (iNOS) which is not constitutively expressed
M Lechleitner et al.
Lipids, 29(9), 627-633 (1994-09-01)
A recently developed immunosuppressive substance, 2-chloro-2-deoxyadenosine (2-CdA), was reported to inhibit monocyte functions at low concentration. Because macrophages play a key role in the formation of atherosclerotic plaques, it was of interest to study the effect of 2-CdA on cellular
J W Huh et al.
Journal of bacteriology, 178(16), 4935-4941 (1996-08-01)
Endogenous ADP-ribosylation was detected in Bacillus subtilis, as determined in vitro with crude cellular extracts. The ADP-ribosylated protein profile changed during growth in sporulation medium, displaying a temporary appearance of two ADP-ribosylated proteins (36 and 58 kDa) shortly after the
M Jesser et al.
Biochemical and biophysical research communications, 195(2), 558-564 (1993-09-15)
A cytoplasmic poly(ADP-ribose)polymerase (PARP) was purified from mouse plasmacytoma free messenger ribonucleoprotein particles using chromatography on 3-aminobenzamide affigel-10. The purified protein showed one band at 116 kDa on SDS-polyacrylamide gel electrophoresis and shared similar antigenic sites to the nuclear PARP.
H Altmann
Acta biologica Hungarica, 41(1-3), 19-34 (1990-01-01)
Poly(ADP-ribose)-polymerase is an important cellular regulatory enzyme which can change chromatin structure and function. Action mechanisms and activation of the enzyme are described. The synthesis of poly(ADP-ribose) can be modulated by interaction of substances with the DNA binding site of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service