Skip to Content
Merck
All Photos(1)

Key Documents

930571

Sigma-Aldrich

Pomalidomide-piperidine-carboxylic acid

≥95%

Synonym(s):

1-[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl], 1-[2-(2,6-Dioxo-3-piperidyl)-1,3-dioxo-isoindolin-5-yl]piperidine-4-carboxylic acid, 4-Piperidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N3O6
CAS Number:
Molecular Weight:
385.37
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21
Pricing and availability is not currently available.

ligand

pomalidomide

Quality Level

Assay

≥95%

form

powder

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(O)C1CCN(C2=CC=C3C(=O)N(C(=O)C3=C2)C4C(=O)NC(=O)CC4)CC1

InChI

1S/C19H19N3O6/c23-15-4-3-14(16(24)20-15)22-17(25)12-2-1-11(9-13(12)18(22)26)21-7-5-10(6-8-21)19(27)28/h1-2,9-10,14H,3-8H2,(H,27,28)(H,20,23,24)

InChI key

LXOQUSNJOGMSJJ-UHFFFAOYSA-N

Application

Pomalidomide-piperidine-carboxylic acid is a functionalized cereblon ligand for development of pomalidomide-based PROTACs. Allows rapid conjugation with amine linkers due to the presence of a carboxyl group via peptide coupling reactions. A basic building block for development of a protein degrader library.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Protein Degrader Building Blocks

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingwei Shao et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(20), e2102555-e2102555 (2021-08-17)
DNA-binding proteins, including transcription factors (TFs), play essential roles in various cellular processes and pathogenesis of diseases, deeming to be potential therapeutic targets. However, these proteins are generally considered undruggable as they lack an enzymatic catalytic site or a ligand-binding
Daniel P Bondeson et al.
Annual review of pharmacology and toxicology, 57, 107-123 (2016-10-13)
Protein homeostasis networks are highly regulated systems responsible for maintaining the health and productivity of cells. Whereas therapeutics have been developed to disrupt protein homeostasis, more recently identified techniques have been used to repurpose homeostatic networks to effect degradation of
Kedra Cyrus et al.
Molecular bioSystems, 7(2), 359-364 (2010-10-06)
Conventional genetic approaches have provided a powerful tool in the study of proteins. However, these techniques often preclude selective manipulation of temporal and spatial protein functions, which is crucial for the investigation of dynamic cellular processes. To overcome these limitations
Philipp M Cromm et al.
Cell chemical biology, 24(9), 1181-1190 (2017-06-27)
Traditional pharmaceutical drug discovery is almost exclusively focused on directly controlling protein activity to cure diseases. Modulators of protein activity, especially inhibitors, are developed and applied at high concentration to achieve maximal effects. Thereby, reduced bioavailability and off-target effects can
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service