Skip to Content
Merck
All Photos(2)

Key Documents

GF35512725

Yttrium

wire reel, 0.05m, diameter 1.0mm, 99.9%

Synonym(s):

Yttrium, Y 005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
MDL number:
UNSPSC Code:
12141617
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 355-127-25

resistivity

57 μΩ-cm, 20°C

L × diam.

0.05 m × 1.0 mm

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Scholz et al.
Dalton transactions (Cambridge, England : 2003), 44(30), 13522-13529 (2015-07-03)
Nanoscopic yttrium acetate fluorides Y(CH(3)COO)(3-z)F(z) and yttrium oxide fluorides YO(3-z)/(2)F(z )were prepared with tunable Y/F molar ratios via the fluorolytic sol-gel route. All samples were characterized by X-ray diffraction, elemental analysis and thermal analysis. In addition, local structures of all
Dan Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4207-4213 (2014-04-18)
YF3:Er3+ hollow nanofibers were successfully fabricated via fluorination of the relevant Y2O3:Er3+ hollow nanofibers which were obtained by calcining the electrospun PVP/[Y(NO3)3 + Er(NO3)3] composite nanofibers. The morphology and properties of the products were investigated in detail by X-ray diffraction
Hojjat Ahmadzadehfar et al.
Seminars in nuclear medicine, 40(2), 105-121 (2010-02-02)
Radioembolization (RE), also termed selective internal radiation therapy (SIRT), has been gradually introduced to the clinical arsenal of cytoreductive modalities in recent years. There is growing evidence for efficiency in liver tumors of various entities, with the most prominent ones
Yoshiko Fujita et al.
Environmental science & technology, 49(16), 9460-9468 (2015-07-02)
Increasing rare earth element (REE) supplies by recycling and expanded ore processing will result in generation of new wastewaters. In some cases, disposal to a sewage treatment plant may be favored, but plant performance must be maintained. To assess the
[Use of lasers on aluminum-yttrium garnet with neodymium in clinical practice].
O K Skobelkin et al.
Khirurgiia, (8)(8), 155-157 (1990-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service