- Nanoscopic yttrium oxide fluorides: non-aqueous fluorolytic sol-gel synthesis and structural insights by 19F and 89Y MAS NMR.
Nanoscopic yttrium oxide fluorides: non-aqueous fluorolytic sol-gel synthesis and structural insights by 19F and 89Y MAS NMR.
Dalton transactions (Cambridge, England : 2003) (2015-07-03)
G Scholz, M Dreger, R Bertram, E Kemnitz
PMID26133504
ABSTRACT
Nanoscopic yttrium acetate fluorides Y(CH(3)COO)(3-z)F(z) and yttrium oxide fluorides YO(3-z)/(2)F(z )were prepared with tunable Y/F molar ratios via the fluorolytic sol-gel route. All samples were characterized by X-ray diffraction, elemental analysis and thermal analysis. In addition, local structures of all samples were studied by (19)F MAS, (19)F-(89)Y CP MAS and (1)H-(89)Y CP MAS NMR spectroscopy and the respective chemical shifts are given. For both classes of compounds, only the fluorination using one equivalent of F (z = 1) leads to defined, well crystalline matrices: yttrium acetate fluoride Y(CH(3)COO)(2)F and r-YOF.
Product Description
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%