Skip to Content
Merck
All Photos(2)

Key Documents

E18425

Sigma-Aldrich

Ethyl cyanoacetate

≥98%

Synonym(s):

(Ethoxycarbonyl)acetonitrile, 2-Cyanoacetic acid ethyl ester, 3-Ethoxy-3-oxopropanenitrile, Cyanoacetic acid ethyl ester, Cyanoacetic ester, Ethyl α-cyanoacetate, Ethyl 2-cyanoacetate, Ethyl cyanacetate, Malonic acid ethyl ester nitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2COOC2H5
CAS Number:
Molecular Weight:
113.11
Beilstein:
605871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 67.8 °C)

Assay

≥98%

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

208-210 °C (lit.)

mp

−22 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC#N

InChI

1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3

InChI key

ZIUSEGSNTOUIPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl cyanoacetate is an ester. Knoevenagel condensation of ethyl cyanoacetate with aldehyde is reported. Microwave enhanced Knoevenegal condensation reaction of ethyl cyanoacetate with an aldehyde, P2O5, piperidine and chlorobenzene is reported.

Ethyl cyanoacetate is widely used as a building block in organic synthesis to produce active pharmaceutical ingredients.

Application

Ethyl cyanoacetate may be used in the synthesis of ethyl glyoxylate. It was used to investigate the Knoevenagel condensation reactions in microreactor using zeolite catalysts obtained by grafting amino groups onto NaX and CsNaX zeolites.

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuya Oka et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(39), 9362-9368 (2017-05-18)
Mesoporous basic Mg-Al mixed metal oxides (MMOs) with a high surface area and large pore size have been prepared through the assembly of monodispersed layered double hydroxide nanoparticles (LDHNPs) with block copolymer templates. The particle sizes of the LDHNPs were
Electrogenerated bases VII. Novel syntheses of ethyl glyoxalate and diethyl ketomalonate via electrogenerated superoxide.
Sugawara M and Baizer MM.
Tetrahedron Letters, 24(22), 2223-2226 (1983)
Microwave Enhanced Knoevenagel Condensation of Ethyl Cyanoacetate with Aldehydes.
Kim S-Y, et al.
Synthetic Communications, 27(4), 533-541 (1997)
Korany A Ali et al.
Mini reviews in medicinal chemistry, 18(8), 717-727 (2017-04-27)
In this study, synthesis, molecular docking and anticancer screening of new series of substituted heterocycles with trimethoxy phenyl scaffold as Combretastatin analogues were described. Substituted pyridines were synthesized via the reaction of (E)-3-(dimethylamino)-1-(3,4,5- trimethoxyphenyl)prop-2-en-1-one (2) with active methylene reagents. Substituted
Mohie E M Zayed et al.
Journal of fluorescence, 27(3), 853-860 (2017-01-23)
4-(2,3,4-trimethoxyphenyl)-8-methoxy-2-oxo-1,2,5,6 tetrahydrobenzo [h] quinoline-3-carbonitrile (TMTQ) dye was synthesized by one-pot multicomponent reactions (MCRs) of 2,3,4 trimethoxybenzaldehyd, ethyl cyanoacetate, 6-methoxy-1,2,3,4-tetrahydro-naphthalin-1-one and ammonium acetate under microwave irradiation. The structures of the synthesized compound was established by spectroscopic (FT-IR

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service