Skip to Content
Merck
All Photos(2)

Key Documents

C88505

Sigma-Aldrich

Cyanoacetic acid

99%

Synonym(s):

2-Cyanoacetic acid, Cyanoethanoic acid, Malonic mononitrile, Monocyanoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2COOH
CAS Number:
Molecular Weight:
85.06
Beilstein:
506325
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.1 mmHg ( 100 °C)

Quality Level

Assay

99%

form

crystals

bp

108 °C/0.15 mmHg (lit.)

mp

64-70 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless to very faintly yellow

SMILES string

OC(=O)CC#N

InChI

1S/C3H3NO2/c4-2-1-3(5)6/h1H2,(H,5,6)

InChI key

MLIREBYILWEBDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyanoacetic acid can be used as a reagent:
  • Along with acetic anhydride for cyanoacetylation of various pyrroles, indoles, and aniline derivatives. It can also be used in other reactions such as cyclizations, syntheses of coumarins and other heterocycles.
  • To prepare of key intermediate via Knoevenagel condensation in the total synthesis of 5-acetamido-substituted melatonin derivatives as MT3 receptor ligands.
  • In the synthesis of aminopyrrolinone derivatives by reacting Ugi adducts of cyanoacetic acid and aromatic aldehydes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of novel ??3 receptor ligands via an unusual Knoevenagel condensation
Volkova MS, et al.
Bioorganic & medicinal chemistry letters, 22(24), 7578-7581 (2012)
Cyanoacetylation of indoles, pyrroles and aromatic amines with the combination cyanoacetic acid and acetic anhydride
Slaett J, et al.
Synthesis, 2004(16), 2760-2765 (2004)
Yijun Huang et al.
Molecular diversity, 15(1), 3-33 (2010-03-02)
The Gewald reaction of sulfur, cyanoacetic acid derivatives, and oxo-component (G-3CR) yielding highly substituted 2-aminothiophene derivatives has seen diverse applications in combinatorial and medicinal chemistry. Its products are of great use in pharmaceutical industry mainly as small molecular weight inhibitors.
B S Kim et al.
Neuroradiology, 45(7), 445-450 (2003-06-05)
Congenital arteriovenous fistulae (AVF) of the internal maxillary artery (IMA) are rare. We present the angiographic findings and management of six AVF of the IMA, selected from 147 patients with facial vascular malformations. The fistula was thought to be congenital
Tian-Yu Liu et al.
Organic & biomolecular chemistry, 4(11), 2097-2099 (2006-05-27)
The bifunctional thiourea-tertiary amine derivatives of simple chiral diamines serve as highly enantioselective catalysts for the Michael addition of alpha-substituted cyanoacetates to vinyl sulfones, giving an efficient protocol for the construction of an all-carbon substituted quaternary stereocentre.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service