Skip to Content
Merck
All Photos(3)

Documents

D73200

Sigma-Aldrich

3,6-Dichloropyridazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2N2
CAS Number:
Molecular Weight:
148.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

65-69 °C (lit.)

SMILES string

Clc1ccc(Cl)nn1

InChI

1S/C4H2Cl2N2/c5-3-1-2-4(6)8-7-3/h1-2H

InChI key

GUSWJGOYDXFJSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yasser M Loksha et al.
Archiv der Pharmazie, 353(3), e1900295-e1900295 (2020-01-17)
Some novel derivatives of 2-alkyl 6-substituted pyridazin-3(2H)-ones were synthesized by condensation of 3,6-dichloropyridazine with the sodium salt of benzyl cyanide, followed by hydrolysis and coupling with alkyl halides. The synthesized compounds were screened as cyclooxygenase (COX)-1/COX-2 inhibitors and as analgesic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service