D129208
Toluene-3,4-dithiol
technical grade, 90%
Synonym(s):
3,4-Dimercaptotoluene, 4-Methyl-1,2-benzenedithiol, ‘Dithiol’
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H3(SH)2
CAS Number:
Molecular Weight:
156.27
Beilstein:
1635270
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
90%
form
powder
bp
135-137 °C/17 mmHg (lit.)
mp
30-33 °C (lit.)
density
1.179 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
Cc1ccc(S)c(S)c1
InChI
1S/C7H8S2/c1-5-2-3-6(8)7(9)4-5/h2-4,8-9H,1H3
InChI key
NIAAGQAEVGMHPM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Used to generate the corresponding dithiolate ligand in a study of dinuclear gold(I) dithiolate complexes.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inorganic Chemistry, 32, 1749-1749 (1993)
Carsten Milsmann et al.
Inorganic chemistry, 48(20), 9754-9766 (2009-09-23)
The reaction of [Ru(III)(cyclam)Cl(2)]Cl with 2 equiv of sodium N,N-diethyldithiocarbamate in methanol afforded [Ru(II)(cyclam)(Et(2)dtc)](BPh(4)) (1(BPh(4))). The same reaction with only 1 equiv of toluene-3,4-dithiol and a base yielded [Ru(cyclam)(tdt)](PF(6)) (2(PF(6))) which was oxidized by 1 equiv of ferrocenium hexafluorophosphate generating
Ulf Ryde et al.
Journal of the American Chemical Society, 132(13), 4512-4513 (2010-03-17)
The active site of the [FeFe] hydrogenases contains two Fe ions bound to one Cys ligand, three CO molecules, two CN(-) ions, and a dithiolate ligand. The nature of the last of these has been much discussed, and it has
Weidong Zhou et al.
Organic letters, 12(18), 4078-4081 (2010-08-18)
A macrocycle and a rotaxane were constructed by virtue of the thiol-yne click reaction under the irradiation of light in high yield, which can proceed at ambient temperature and humidity under an air atmosphere. Two disulfide macrocycles were synthesized through
Kazunari Yoshizawa
Accounts of chemical research, 45(9), 1612-1621 (2012-06-16)
The transfer of electrons in molecules and solids is an essential process both in biological systems and in electronic devices. Devices that take advantage of the unique electronic properties of a single molecule have attracted much attention, and applications of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service