Skip to Content
Merck
All Photos(2)

Documents

763969

Sigma-Aldrich

Benzene-1,4-dithiol

99% (GC)

Synonym(s):

1,4-Benzenedithiol, 1,4-Dimercaptobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6S2
CAS Number:
Molecular Weight:
142.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

form

powder or crystals

mp

92-97 °C

storage temp.

2-8°C

SMILES string

Sc1ccc(S)cc1

InChI

1S/C6H6S2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

WYLQRHZSKIDFEP-UHFFFAOYSA-N

General description

Benzene-1,4-dithiol (BDT) is a class of aromatic dithiol that can be used to form a self-assembled monolayer (SAM) on a variety of surfaces.

Application

BDT can be used as a cross-linker that facilitates the development of acrylate based light sensitive epoxidized soya bean oil for 3D printing applications. Gold based nanoconstrictions can be fabricated by functionalizing the surface with BDT, which reduces the overall flicker noise with respect to the unmodified surface.
This molecule can be used in single molecule junctions in electronic devices. It has also shown use in nanomaterial synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Lin et al.
ACS nano, 12(7), 6492-6503 (2018-06-21)
Charge transport plays an important role in defining both far-field and near-field optical response of a plasmonic nanostructure with an ultrasmall built-in nanogap. As the gap size of a gold core-shell nanomatryoshka approaches the sub-nanometer length scale, charge transport may
Photoresins based on acrylated epoxidized soybean oil and benzenedithiols for optical 3D printing
Miezinyte G, et al.
Rapid Prototyping Journal (2018)
Conductance of a molecular junction
Reed MA, et al.
Science, 278(5336), 252-254 (1997)
Noise spectroscopy of tunable nanoconstrictions: molecule-free and molecule-modified
Handziuk V, et al.
Nanotechnology, 29(38), 385704-385704 (2018)
Yung-Ching Chang et al.
Optics express, 25(20), 24767-24779 (2017-10-19)
Near-field and far-field optical properties of plasmonic materials can be tailored by coupling the existing structures. However, fabricating 3D coupled structures in the solution by molecular linkers may suffer from low yield, low stability (particle aggregates), long reaction time, complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service