CLS1395250
Pyrex® round media storage bottles and reusable screw caps
capacity 250 mL
Synonym(s):
Pyrex® media bottles, bottle, glass media bottles, laboratory bottles, media bottle, pyrex bottle, round bottles
About This Item
Recommended Products
material
borosilicate glass bottle
clear bottle
cylindrical bottle
orange polypropylene cap (plug seal)
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
graduations
reusable
packaging
case of 10
manufacturer/tradename
Corning 1395-250
availability
not available in EU
capacity
250 mL
diam. × H
70 mm × 138 mm
fitting
GL 45 neck thread
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Pyrex round media storage bottles are made of borosilicate glass with permanent, white enamel graduations and marking spots.
Caution: Bottles larger than 2 L should NOT be used with bottle-top filter units or in other applications involving vacuum/pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.
A wide range of optional caps are available for these bottles:
- Vented membrane cap with 0.22 μm PTFE hydrophobic membrane
- Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C)
- PBT caps with silicone septa for syringe sampling or introduction of reagents
Caution: DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. -A wide range of optional caps are available for these bottles: -Polypropylene colored caps in a choice of orange, green, purple, red, and light gray for ease of sorting and identification. -Vented membrane cap with 0.22 μm PTFE hydrophobic membrane. -Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C). -PBT caps with silicone septa for syringe sampling or introduction of reagents.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service