Key Documents
BR122710
BRAND® Media Bottle
capacity 500 mL, borosilicate glass 3.3 bottle, polypropylene cap, pack of 4 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass 3.3 bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL56)
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 4 ea
manufacturer/tradename
BRAND 122710
capacity
500 mL
General description
The BRAND® Media Bottle is expertly crafted from Boro 3.3 glass, ensuring excellent durability and resistance to thermal and chemical stress. Designed with a GL 56 neck, this bottle features a low height for easy handling and storage. The PP screw-on cap provides a secure, vacuum-tight seal, making it ideal for sterile filtration applications. The bottle is also equipped with a clear scale for accurate volume measurement, enhancing its usability in laboratory settings. This versatile media bottle is essential for storing and handling various media and solutions, ensuring reliability and safety in your experiments.
Features and Benefits
  • Bottle and cap made of biologically inert material
  • Larger bottle neck and bottle can be tilted by 45° for easy removal of medium
  • Compatible with bottle top filters
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)