Skip to Content
Merck
All Photos(1)

Documents

B74607

Sigma-Aldrich

1-Bromononane

98%

Synonym(s):

Nonyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8Br
CAS Number:
Molecular Weight:
207.15
Beilstein:
1733632
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

201 °C (lit.)

density

1.084 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCBr

InChI

1S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3

InChI key

AYMUQTNXKPEMLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Balgavý et al.
General physiology and biophysics, 5(4), 365-370 (1986-08-01)
Interaction of chemical fusogen n-nonyl bromide with a model membrane formed from phosphatidylcholine was studied using 2D-NMR spectra of heavy water and 31P-NMR proton decoupled spectra of the lipid phosphate group in multilamellar lipid dispersions. n-Nonyl bromide was found to
Hypomethylating agents (HMAs) effect on myelodysplastic/myeloproliferative neoplasm unclassifiable (MDS/MPN-U): single institution experience.
Aref Al-Kali et al.
Leukemia & lymphoma, 1-3 (2018-02-22)
Anna Parus et al.
Ecotoxicology and environmental safety, 208, 111595-111595 (2021-01-06)
Modern agricultural practices are often based on the use of mixtures of specific herbicides to achieve efficient crop protection. The major drawbacks of commercial herbicidal formulations include the necessity to incorporate toxic surfactants and high volatility of active substances. Transformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service