Skip to Content
Merck
All Photos(2)

Key Documents

935727

Sigma-Aldrich

Barium hydroxide octahydrate

greener alternative

≥99.9% trace metals basis

Synonym(s):

Barium dihydroxide octahydrate, Dihydroxybarium octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · 8H2O
CAS Number:
Molecular Weight:
315.46
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.21

Quality Level

Assay

≥99.9% trace metals basis

form

crystals, granules or powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

mp

78 °C (lit.)

greener alternative category

SMILES string

O.O.O.O.O.O.O.O.O[Ba]O

InChI

1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2

InChI key

ZUDYPQRUOYEARG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium hydroxide crystalizes as octahydrate. At 100°C, it transforms into barium hydroxide monohydrate, which produces barium oxide and water. It serves as a precursor in the synthesis of a compound containing barium. It is also employed in the titration of weak acids. Furthermore, the hydrolysis of ester and nitriles uses barium hydroxide octahydrate as a powerful base
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Barium hydroxide octahydrate is used as a catalyst for the synthesis of methyl ester from rubber seed oil For thermal storage, barium hydroxide octahydrate is an important phase change material with a low melting point. It has a large heat capacity, as well as supercooling and phase separation phenomena during the cooling process Additionally, barium hydroxide octahydrate was used to produce a sodium gold sulfite solution, which was later used for electroless plating of thin gold films directly onto silicon nitride thin films and into micropores
Catalyzes the β-elimination of phosphoserine residues.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental investigation of barium hydroxide octahydrate as latent heat storage materials
Wang Q, et al.
Solar Energy, 177, 99-107 (2019)
NAPHTHORESORCINOL
Meyer K, et al.
Organic Syntheses, 3, 637-637 (1945)
Electroless plating of thin gold films directly onto silicon nitride thin films and into micropores
Whelan J C, et al.
ACS Applied Materials & Interfaces, 6, 10952?10957-10952?10957 (2014)
Two-step process for production of methyl ester from rubber seed oil using barium hydroxide octahydrate catalyst: Process optimization
Reshad A, et al.
Journal of Cleaner Production, 142, 3490?3499-3490?3499 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service