450146
Calcium hydroxide
99.995% trace metals basis
Synonym(s):
Calcium dihydroxide, Carboxide
About This Item
Recommended Products
Quality Level
Assay
99.995% trace metals basis
form
powder
impurities
<3% calcium carbonate
density
2.24 g/mL at 25 °C (lit.)
SMILES string
O[Ca]O
InChI
1S/Ca.2H2O/h;2*1H2/q+2;;/p-2
InChI key
AXCZMVOFGPJBDE-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service