931012
9,9′-(5-(4,6-Diphenyl-1,3,5-triazin-2-yl)-1,3-phenylene)bis(9H-carbazole)
≥97% (HPLC)
Synonym(s):
DCzTrz
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C45H29N5
CAS Number:
Molecular Weight:
639.75
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
grade:
sublimed grade
solubility:
chloroform: soluble
toluene: soluble
toluene: soluble
Recommended Products
grade
sublimed grade
Quality Level
Assay
≥97% (HPLC)
energy gap
0.25 eV
loss
0.5% TGA, >300 °C (weight loss)
solubility
chloroform: soluble
toluene: soluble
fluorescence
λem 449 nm±10 nm in toluene
Orbital energy
HOMO 5.88 eV
LUMO 2.86 eV
λ
in toluene
UV absorption
λ: 337 nm±5 nm Amax
Related Categories
Application
DCzTrz can be used in studies on organic light-emitting diodes (OLEDs). It can be used as an electron-transport layer, facilitating the efficient injection and transport of negative charges (electrons) from the cathode to the emissive layer. DCzTrz has been studied for its potential use as an electron-transport material to assist in the extraction and collection of negative charges generated in the active layer in organic photovoltaic (OPV) devices.
Use as OLED Materials: TADF Materials, Blue Dopant
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mounggon Kim et al.
Advanced materials (Deerfield Beach, Fla.), 27(15), 2515-2520 (2015-03-11)
High quantum efficiency above 18% and extended lifetime three times longer than that of phosphorescent organic light-emitting diodes (OLEDs) are demonstrated in blue thermally activated delayed fluorescent OLEDs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service