Skip to Content
Merck
All Photos(2)

Key Documents

932329

Sigma-Aldrich

DMAC-TRZ

≥99% (HPLC)

Synonym(s):

10-(4-(4,6-Diphenyl-1,3,5-triazin-2-yl)phenyl)-9,9-dimethyl-9,10-dihydroacridine, 9,10-Dihydro-9,9-dimethyl-10-(4-(4,6-diphenyl-1,3,5-triazin-2-yl)phenyl)acridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H28N4
CAS Number:
Molecular Weight:
516.63
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥99% (HPLC)
grade:
sublimed grade
solubility:
toluene: soluble

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

energy gap

0.05 eV (delta EST)

description

2.52 eV (HOMO-LUMO gap)

transition temp

Tg 91 °C

solubility

toluene: soluble

fluorescence

λem 495 nm±10 nm in toluene (PL)

Orbital energy

HOMO - 5.30 eV 
LUMO - 2.78 eV 

λ

in toluene

UV absorption

λ: 390 nm±20 nm Amax

Application

DMAC-TRZ can be used in R&D studies such as organic light-emitting diodes (OLEDs), organic photovoltaic (OPV) devices, organic field-effect transistors (OFETs) and Dye-sensitized solar cells (DSSCs). DMAC-TRZ has energy levels that align well with other materials commonly used in electronic devices. This property allows for efficient charge injection, extraction and transport across different layers, which contributes to improved device operation and performance.
Sublimed high purity material used as TADF Blue-green Dopant, with EQE of 26.5%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A versatile thermally activated delayed fluorescence emitter for both highly efficient doped and non-?doped organic light emitting devices
Tsai Wei-Lung, et al.
Chemical Communications (Cambridge, England), 51, 13662-13662 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service