Skip to Content
Merck
All Photos(2)

Key Documents

930938

Sigma-Aldrich

Lithium nitrate

greener alternative

battery grade, 99.999% trace metals basis

Synonym(s):

Lithium salt of nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

grade

battery grade

Quality Level

Assay

99.999% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤15 ppm (trace metals analysis)

mp

264 °C (lit.)

solubility

soluble (H2O: highly soluble(lit.); alcohols: soluble(lit.); acetone: soluble(lit.))

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[O-][N+]([O-])=O

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1

InChI key

IIPYXGDZVMZOAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium nitrate is a white, crystalline salt that is soluble in water, ethanol, methanol, pyridine, ammonia, and acetone. Importantly, it is also highly soluble up to 5 wt% in ether-based solvents such as dimethoxyethane (DME) and 1,3-dioxolane (DOL), but only soluble up to 1 wt% in carbonate-based solvents like ethylene carbonate (EC) and diethtyl carbonate (DEC).
Lithium nitrate is produced by reacting nitric acid and lithium carbonate, which evolves carbon dioxide and water. The resulting material is purified and dried.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Researchers and manufacturers use lithium nitrate in the synthesis of many lithium compounds. Our 99.999% lithium nitrate is well-suited as a reagent for solid-state syntheses of lithium metal oxides, especially where purity is of high importance, for example, when making products whose fundamental properties are under investigation.
Our 99.999% lithium nitrate is also well-suited for use as an additive to electrolytes in lithium-sulfur batteries and lithium metal batteries. Lithium nitrate can passivate the surface of lithium metal and suppress the redox shuttle of the dissolved lithium polysulfides on the lithium anode. In one study, the addition of 0.3 M LiNO3 nearly doubled the gravimetric capacity of lithium-sulfide batteries. Another study found that the dissolution of 1 to 5 wt% LiNO3 to the electrolyte suppressed growth of lithium dendrites and extended cycle lifetimes. Similarly beneficial effects of lithium nitrate as an additive have been observed with Li2S cathodes, carbon nanofiber-encapsulated sulfur cathodes, cobalt sulfide (Co3S4) cathodes, and polyacrylonitrile-sulfur composite cathodes. Even lithium metal anodes with LiNi0.8Co0.15Al0.05O2 (NCA) cathodes with LiNO3 added to the electrolyte showed higher coulombic efficiencies and suppressed dendrite formation compared to the electrolyte without LiNO3.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of LiNO3 in rechargeable lithium/sulfur battery.
Zhang S, et al.
Electrochimica Acta, 70, 344-348 (2012)
Yuan Yang et al.
Journal of the American Chemical Society, 134(37), 15387-15394 (2012-08-23)
Li(2)S is a high-capacity cathode material for lithium metal-free rechargeable batteries. It has a theoretical capacity of 1166 mAh/g, which is nearly 1 order of magnitude higher than traditional metal oxides/phosphates cathodes. However, Li(2)S is usually considered to be electrochemically
Weiyang Li et al.
Nature communications, 6, 7436-7436 (2015-06-18)
Lithium metal has shown great promise as an anode material for high-energy storage systems, owing to its high theoretical specific capacity and low negative electrochemical potential. Unfortunately, uncontrolled dendritic and mossy lithium growth, as well as electrolyte decomposition inherent in
Shuya Wei et al.
Journal of the American Chemical Society, 137(37), 12143-12152 (2015-09-02)
Sulfur/polyacrylonitrile composites provide a promising route toward cathode materials that overcome multiple, stubborn technical barriers to high-energy, rechargeable lithium-sulfur (Li-S) cells. Using a facile thermal synthesis procedure in which sulfur and polyacrylonitrile (PAN) are the only reactants, we create a
Tao Chen et al.
Journal of the American Chemical Society, 139(36), 12710-12715 (2017-08-25)
Lithium-sulfur batteries (Li-S) have attracted soaring attention due to the particularly high energy density for advanced energy storage system. However, the practical application of Li-S batteries still faces multiple challenges, including the shuttle effect of intermediate polysulfides, the low conductivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service