Skip to Content
Merck
All Photos(2)

Key Documents

229741

Sigma-Aldrich

Lithium nitrate

99.99% trace metals basis

Synonym(s):

Lithium salt of nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
99.99% trace metals basis
form:
crystalline powder
solubility:
522 g/L at 20 °C

Quality Level

Assay

99.99% trace metals basis

form

crystalline powder

mp

264 °C (lit.)

solubility

522 g/L at 20 °C

application(s)

battery manufacturing

functional group

nitrate

SMILES string

[Li+].[O-][N+]([O-])=O

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1

InChI key

IIPYXGDZVMZOAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium nitrate has been used as an additive to the electrolyte solution of bis(trifluoromethane)sulfonimide in lithium-sulfur batteries to increase the coulombic efficiency. It can also be used in the sol-gel synthesis of spinel lithium titanate (Li4Ti5O12) materials.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balancing the chemisorption and charge transport properties of the interlayer in lithium-sulfur batteries
Yang L, et al.
Journal of Material Chemistry A, 5(24), 12506-12512 (2017)
Ji-Yoon Song et al.
Nanomaterials (Basel, Switzerland), 8(2) (2018-02-08)
For practical application of lithium-sulfur batteries (LSBs), it is crucial to develop sulfur cathodes with high areal capacity and cycle stability in a simple and inexpensive manner. In this study, a carbon cloth infiltrated with a sulfur-containing electrolyte solution (CC-S)
Separator for lithium-sulfur battery based on polymer blend membrane
Freitag A, et al.
Journal of Power Sources, 363, 384-391 (2017)
Sol-Gel Synthesis of Nanocomposite Cu-Li4Ti5O12 Structures for Ultrahigh Rate Li-Ion Batteries
Erdas A, et al.
Acta Physica Polonica A, 127(24), 1026-1026 (2015)
Operando characterization of intermediates produced in a lithium-sulfur battery
Gorlin Y, et al.
Journal of the Electrochemical Society, 162(7), A1146-A1155 (2015)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service