Skip to Content
Merck
All Photos(1)

Key Documents

925756

Sigma-Aldrich

N-([1,1′-Biphenyl]-4-yl)-9,9-dimethyl-N-(4-(9-phenyl-9H-carbazol-3-yl)phenyl)-9H-fluoren-2-amine

≥97%

Synonym(s):

BCFN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H38N2
CAS Number:
Molecular Weight:
678.86
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

description

PL: 413 nm (in DCM)

Quality Level

Assay

≥97%

form

powder

color

white to yellow

Orbital energy

HOMO 5.3 eV 
LUMO 3.2 eV 

λ

in dichloromethane

UV absorption

λ: 353 nm Amax

SMILES string

[n]2(c3c(c%10c2cccc%10)cc(cc3)c4ccc(cc4)N(c8ccc(cc8)c9ccccc9)c5cc6c(cc5)c7c(cccc7)C6(C)C)c1ccccc1

InChI

1S/C51H38N2/c1-51(2)47-19-11-9-17-43(47)44-31-30-42(34-48(44)51)52(40-26-21-36(22-27-40)35-13-5-3-6-14-35)41-28-23-37(24-29-41)38-25-32-50-46(33-38)45-18-10-12-20-49(45)53(50)39-15-7-4-8-16-39/h3-34H,1-2H3

InChI key

GJWBRYKOJMOBHH-UHFFFAOYSA-N

Application

N-([1,1′-biphenyl]-4-yl)-9,9-dimethyl-N-(4-(9-phenyl-9H-carbazol-3-yl)phenyl)-9H-fluoren-2-amine (BCFN) is an electron-rich material, with the HOMO energy level of 5.3 eV, the LUMO energy level of 2.2 eV and high hole mobility of 5.43 × 10-5 cm2V-1s-1. As a result, it is used as a hole-transport and hole-injection material in organic light-emitting diodes. BCFN has shown its advantages in improving the hole mobility of polymer-based hole transport layer; lower driving voltage of device, better efficiency, and higher EQE due to exciplex formation by utilizing in EML; and better device lifetimes in the combination of Ir(piq)2(acac) to act as an electron scavenger layer for an reported device.

BCFN is a good morphologically stable material with Tg of 125 °C and Td of 375 °C.

With a ET of 2.58 eV, BCFN is also a very good charge transporting layer material due to its large band gap (Eg = 3.1 eV) 2 for green, red and white phosphorescent organic light-emitting diodes (PhOLEDs).

OLED Device Performance:
BCFN increase the hole transport mobility of PVK (pure PVK: 2.09×10-9 V cm-1 S-1; 30% of BCFN in PVK: 9.86×10-7 V cm-1 S-1; 50% of BCFN in PVK: 1.15×10-5 V cm-1 S-1):
1. ITO/PEDOT:PSS (40 nm)/PVK:BCFN (3:7, 20 nm)/BCFN:PFO (x:1-x, 80 nm)/CsF (1.5 nm)/Al (120 nm)
Color: Blue


BCFN in combination of ET1 to act as EML in forming exciplex and improve device performance:
2. ITO/HIL/HTL/(ET1+P1) or (ET1+BCFN+P1(doped)/ETL/ LiF/Al
Color: red
Device emission area of 81cm2, and at 1000 cd m-2, the efficiency exceeds 200 lm/W and the EQE exceeds 50 %.
lifespan@95: from 71h to 133h

BCFN in combination with Ir(piq)2(acac) as electron scavenger layer:
3. ITO/NDP series in BCFN (3wt.%, 10nm)/BCFN(160nm) or BCFN(130nm)+BCFN:Ir(piq)2(acac)(2wt.%, 10nm)+BCFN(20)/ [Ir(TMSBppy)ppy2]:BPTZBC (10 wt.%, 40nm))/ TPBi:Liq (1:1 wt. % ratio, 36nm)/ Liq (0.1nm)/Al (60 nm)
Color: yellow
Efficiencies are not affected.
lifespan@95: from 71h to 133h
N-([1,1′-Biphenyl]-4-yl)-9,9-dimethyl-N-(4-(9-phenyl-9H-carbazol-3-yl)phenyl)-9H-fluoren-2-amine can be used in different application areas as an emissive layer or hole transport layer due the presence of π-conjugated structure and electron-donating/accepting groups. It is suitable to be used in R&D studies such as organic field-effect transistors (OFETs), molecular electronics, sensors, organic photovoltaics (OPVs), and organic light-emitting diodes (OLEDs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient blue polyfluorenes using blending materials as hole transport layer.
Liang J, et al
Organic Electronics, 51, 111-118 (2017)
Achieving highly efficient blue light-emitting polymers by incorporating a styrylarylene amine unit.
Peng F, et al.
Journal of Material Chemistry C, 6, 12355-12363 (2018)
Improved efficiency of blue polymer light-emitting diodes using a hole transport material.
Liang, J, et al.
Journal of Material Chemistry C, 5, 5096-5101 (2017)
Highly efficient single-unit white OLED device with emission from both singlet and triplet excitons
Ishisone,T., et al.
Society for Information Display International Symposium Digest of Technical Papers, 22, 404-411 (2015)
Boron-based TADF emitters with improved OLED device efficiency roll-off and long lifetime.
Bell B M, et al.
Dyes and Pigments, 141, 83-92 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service