Skip to Content
Merck
All Photos(2)

Key Documents

901494

Sigma-Aldrich

N-Methylated pomalidomide

≥98%

Synonym(s):

4-Amino-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione, E3 Ligase ligand methylated negative control, Ligand for PROTAC® research

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3O4
CAS Number:
Molecular Weight:
287.27
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

ligand

N-Methylated Pomalidomide

Quality Level

Assay

≥98%

form

powder or crystals

reaction suitability

reagent type: ligand

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O=C(C(CC1)N(C2=O)C(C3=C2C=CC=C3N)=O)N(C)C1=O

InChI

1S/C14H13N3O4/c1-16-10(18)6-5-9(13(16)20)17-12(19)7-3-2-4-8(15)11(7)14(17)21/h2-4,9H,5-6,15H2,1H3

InChI key

YSWQOCGODHPKED-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
901487901490923931
reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

powder or crystals

form

powder or crystals

form

powder or crystals

form

-

ligand

N-Methylated Pomalidomide

ligand

VH032

ligand

VH032

ligand

pomalidomide

Application

N-Methylated pomalidomide is a ligand used as a negative control for pomalidomide (P0018) in the recruitment of the Cereblon (CRBN) protein for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Lu et al.
Chemistry & biology, 22(6), 755-763 (2015-06-09)
BRD4, a bromodomain and extraterminal domain (BET) family member, is an attractive target in multiple pathological settings, particularly cancer. While BRD4 inhibitors have shown some promise in MYC-driven malignancies such as Burkitt's lymphoma (BL), we show that BRD4 inhibitors lead to
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service