Skip to Content
Merck
All Photos(1)

Key Documents

855642

Sigma-Aldrich

N-Acetyl-L-glutamic acid

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

(2S)-2-Acetamidopentanedioic acid, N-Acetylglutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
189.17
Beilstein:
1727473
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-Acetyl-L-glutamic acid, ReagentPlus®, 99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

optical activity

[α]22/D −15.6°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

194-196 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m0/s1

InChI key

RFMMMVDNIPUKGG-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiguang Wang et al.
Poultry science, 99(7), 3583-3593 (2020-07-04)
The present study determined the effects of in ovo feeding (IOF) of N-acetyl-L-glutamate (NAG) on early intestinal development and growth performance of broilers. A total of 702 fertile broiler eggs were randomly divided into 3 treatments: 1) non-punctured control group
M Tuchman et al.
Pediatric research, 27(4 Pt 1), 408-412 (1990-04-01)
N-acetylglutamate (NAG) content was measured in homogenates of liver and small intestine obtained from normal and 24-h starved syngeneic mice. Subsequently, NAG was determined in normal, and in carbamyl phosphate synthetase I and ornithine transcarbamylase enzyme-deficient human liver tissue homogenates.
W Zhang et al.
Clinical and investigative medicine. Medecine clinique et experimentale, 13(4), 183-188 (1990-08-01)
Liver tissue from 3 patients with carbamyl phosphate synthetase I deficiency and 11 patients with ornithine transcarbamylase deficiency was analyzed by Western blots for the presence of carbamyl phosphate synthetase I and ornithine transcarbamylase cross-reactive material with anti-rat enzyme antibodies.
J Vockley et al.
Biochemical medicine and metabolic biology, 47(1), 38-46 (1992-02-01)
N-Acetyl-L-glutamate synthetase (NAG synthetase) is a mitochondrial matrix enzyme which catalyzes the synthesis of N-acetyl-Lglutamate (NAG), a physiologic activator of the urea cycle enzyme carbamylphosphate synthetase I. Deficiency of NAG synthetase in humans has been reported only three times previously.
K Sugahara et al.
Journal of chromatography. B, Biomedical applications, 657(1), 15-21 (1994-07-01)
Liquid chromatography-atmospheric pressure chemical-ionization mass spectrometry (LC-APCI-MS) was used for the analysis of N-acetylamino acids that could not be determined with an amino acid analyzer. LC-APCI-MS could directly detect the protonated molecular ions of various synthetic N-acetylamino acids, distinguishing N-acetylserine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service