Skip to Content
Merck
All Photos(2)

Key Documents

851590

Sigma-Aldrich

L-(−)-Xylose, mixture of anomers

≥99%

Synonym(s):

L-Xylose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723080
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

powder

optical activity

[α]24/D −18.7°, c = 4 in H2O

mp

150-152 °C (lit.)

SMILES string

O[C@H]1COC(O)[C@@H](O)[C@@H]1O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5+/m1/s1

InChI key

PYMYPHUHKUWMLA-WISUUJSJSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Misun Lee et al.
Biotechnology for biofuels, 13, 5-5 (2020-01-16)
Efficient bioethanol production from hemicellulose feedstocks by Saccharomyces cerevisiae requires xylose utilization. Whereas S. cerevisiae does not metabolize xylose, engineered strains that express xylose isomerase can metabolize xylose by converting it to xylulose. For this, the type II xylose isomerase
Kyle V Probst et al.
Biotechnology progress, 33(4), 1096-1103 (2017-04-04)
Cyclopentyl methyl ether (CPME) was evaluated for extracting oil or triacylglycerol (TAG) from wet cells of the oleaginous yeast Lipomyces starkeyi. CPME is a greener alternative to chloroform as a potential solvent for oil recovery. A monophasic system of CPME
Yang Liu et al.
Antonie van Leeuwenhoek, 109(2), 207-213 (2015-11-22)
Four Gram-stain positive bacterial strains, designated as 4R1(T), 4R9, 4L13 and 4L18, isolated from seeds of hybrid maize (Zea mays L., Jingke 968), were investigated using a polyphasic taxonomic approach. The cells were found to be facultatively aerobic, motile, spore-forming
Julian Quehenberger et al.
International journal of molecular sciences, 20(1) (2019-01-10)
While in search of an enzyme for the conversion of xylose to xylitol at elevated temperatures, a xylose reductase (XR) gene was identified in the genome of the thermophilic fungus Chaetomium thermophilum. The gene was heterologously expressed in Escherichia coli
Fayin Zhu et al.
Journal of industrial microbiology & biotechnology, 47(2), 223-232 (2020-01-29)
It is of great economic interest to produce succinate from low-grade carbon sources, e.g., lignocellulosic biomass hydrolysate, which mainly contains glucose and xylose. Inactivation of the glucose uptake system PtsG was evaluated for succinate production from xylose-rich feedstocks. Strains with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service