Skip to Content
Merck
All Photos(1)

Key Documents

81365

Sigma-Aldrich

Mowiol® 18-88

Mw ~130,000

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

crystals

mol wt

Mw ~130,000

extent of labeling

86.7-88.7 mol% hydrolysis
~2,700 polymerization

ign. residue

≤0.5%

viscosity

16-20 mPa.s, 4 % in H2O(20 °C)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mowiol®18-88 is a polyvinyl alcohol (PVA) that is mainly used as a surfactant. It can be used as a polymeric matrix in the preparation of silver nanoparticles.

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photo-triggered release in polyamide nanosized capsules
AIP Conference Proceedings, 1599(1), 234-237 (2014)
Influence of polymeric surfactants on pesticidal suspension concentrates: dispersing ability, milling efficiency and stabilization power
Haas S, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 183(1), 785-793 (2001)
Photochemical synthesis of silver nanoparticles in polyvinyl alcohol matrices
Kutsenko AS and Granchak VM
Theoretical and Experimental Chemistry, 45(5), 313-318 (2009)
Roger F Falkenstern-Ge et al.
Journal of bronchology & interventional pulmonology, 20(1), 58-62 (2013-01-19)
Emphysema is a progressive and irreversible disease for which there is no cure to date. The patients experience debilitating shortness of breath with repetitive exacerbations and poor quality of daily life. At present, patients with severe emphysema have limited treatment
Cijun Shuai et al.
Biofabrication, 5(1), 015014-015014 (2013-02-07)
A tetragonal polyvinyl alcohol (PVA) scaffold with 3D orthogonal periodic porous architecture was fabricated via selective laser sintering (SLS) technology. The scaffold was fabricated under the laser power of 8 W, scan speed of 600 mm min(-1), laser spot diameter of 0.8 mm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service