Skip to Content
Merck
All Photos(1)

Documents

324590

Sigma-Aldrich

Mowiol® 40-88

average Mw ~205,000 g/mol

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

crystals

mol wt

average Mw ~205,000 g/mol

mp

>300 °C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mowiol® 40-88 can be used for a variety of applications such as drug delivery, surface enhanced Raman spectroscopy (SERS) fiber-optic sensors, and transparent conducting oxides (TCOs).

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-temperature processing of transparent conductive indium tin oxide nanocomposites using polyvinyl derivatives
Maksimenko I and Wellmann PJ
Thin Solid Films, 520(4), 1341-1347 (2011)
Swelling-activated drug delivery systems
Conte U, et al.
Biomaterials, 9(6), 489-493 (1988)
Fiber-optic SERS sensor with optimized geometry: testing and optimization
Pesapane A, et al.
Journal of Raman Spectroscopy, 41(3), 256-267 (2010)
Igor Bryukhovetskiy et al.
Oncology letters, 13(2), 738-746 (2017-03-31)
Glioblastoma multiforme is an invasive malignant glial brain tumor with a poor prognosis for patients. The primary reasons that lead to the development of treatment resistance are associated with tumor cells infiltrating the brain parenchyma and the specific properties of
Marti Quevedo et al.
Nature communications, 10(1), 2669-2669 (2019-06-19)
The Mediator complex regulates transcription by connecting enhancers to promoters. High Mediator binding density defines super enhancers, which regulate cell-identity genes and oncogenes. Protein interactions of Mediator may explain its role in these processes but have not been identified comprehensively.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service