Skip to Content
Merck
All Photos(1)

Key Documents

776025

Sigma-Aldrich

Maleimidolinourea-Rh110-trimethyl lock

95%

Synonym(s):

Maleimidourea-rhodamine-110 trimethyl lock

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H40N4O9
CAS Number:
Molecular Weight:
756.80
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

storage temp.

−20°C

SMILES string

O=C(O1)C2=CC=CC=C2C31C(C=CC(NC(NCCCN4C(C=CC4=O)=O)=O)=C5)=C5OC6=C3C=CC(NC(CC(C)(C)C7=C(OC(C)=O)C=C(C)C=C7C)=O)=C6

InChI

1S/C43H40N4O9/c1-24-19-25(2)39(35(20-24)54-26(3)48)42(4,5)23-36(49)45-27-11-13-31-33(21-27)55-34-22-28(46-41(53)44-17-8-18-47-37(50)15-16-38(47)51)12-14-32(34)43(31)30-10-7-6-9-29(30)40(52)56-43/h6-7,9-16,19-22H,8,17-18,23H2,1-5H3,(H,45,49)(H2,44,46,53)

InChI key

XMGVLBGRSCGJDI-UHFFFAOYSA-N

Application

Maleimidourea-rhodamine-110 trimethyl lock may be used:
  • Rhodamine 110 based pro-fluorophore activated by esterases in cells.
  • Maleimide functionalized for conjugation to thiolates of small molecules or biomolecules.
  • High signal to noise ratio compared to fluorescein diacetate.
  • Able to fluorescently monitor passage of biomolecules into cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke D Lavis et al.
ACS chemical biology, 1(4), 252-260 (2006-12-14)
Traditional small-molecule fluorophores are always fluorescent. This attribute can obscure valuable information in biological experiments. Here, we report on a versatile "latent" fluorophore that overcomes this limitation. At the core of the latent fluorophore is a derivative of rhodamine in
Sunil S Chandran et al.
Journal of the American Chemical Society, 127(6), 1652-1653 (2005-02-11)
Fluorescent molecules are essential for basic research in the biological sciences and have numerous practical applications. Herein is described the synthesis and use of a new class of latent fluorophores based on a novel design element, the trimethyl lock, that

Related Content

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service