Skip to Content
Merck
All Photos(2)

Key Documents

MABN328

Sigma-Aldrich

Anti-peptidyl-citrulline, clone F95 Antibody

clone F95, from mouse

Synonym(s):

Peptidyl-citrulline, Protein-bound citrulline

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

F95, monoclonal

species reactivity

mouse, rat, chicken, human

species reactivity (predicted by homology)

all (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

shipped in

wet ice

target post-translational modification

unmodified

General description

Deimination (citrullination) is a post translational modification in which the peptidyl-arginine deiminase family of enzymes convert the guanidino side chain of the amino acid arginine into a ureido group, yeilding peptidyl-citrulline. As a result, anti-peptidyl-citrulline antibodies should also cross react with peptidyl-homocitrulline, produced by the carbamylation of lysine, but not free citrulline, which is created in the urea cycle, or as a by-product of nitric oxide synthesis.

Immunogen

Citrullinated peptide consisting of 10 citrulline residues and a carboxyl Gly-Gly-Cys.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected peptidyl-citrulline in human cerebellum, and rat cerebellum tissue.
Western Blotting Analysis: A representative lot detected peptidyl-citrulline in CNS proteins from brains and spinal cord of EAE mice (Nicholas et al., 2005).
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-peptidyl-citrulline, clone F95 Antibody is validated for use in Immunohistochemistry and Western Blotting and Immunocytochemistry for the detection of peptidyl-citrulline.

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Imunnohistochemistry Analysis: A 1:250 dilution of this antibody detected peptidyl-citrulline inhuman cerebral cortex tissue.

Target description

Various kDa calculated

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael F Criscitiello et al.
International journal of molecular sciences, 21(8) (2020-04-25)
The bovine immune system is known for its unusual traits relating to immunoglobulin and antiviral responses. Peptidylarginine deiminases (PADs) are phylogenetically conserved enzymes that cause post-translational deimination, contributing to protein moonlighting in health and disease. PADs also regulate extracellular vesicle
R Bright et al.
Journal of periodontal research, 53(1), 139-143 (2017-10-19)
Postranslational modification of proteins can lead to the production of autoantibodies and loss of immune tolerance. This process has been hypothesised to be a critical factor in the pathogenesis of rheumatoid arthritis. The objective of this study was to demonstrate
Sanna Turunen et al.
Arthritis research & therapy, 18(1), 239-239 (2016-10-22)
Seropositive rheumatoid arthritis (RA) is characterized by autoantibodies binding to citrullinated and homocitrullinated proteins. We wanted to study the expression patterns of these disease-associated protein forms and if the rheumatoid nodule and synovial tissue itself contain biologically active levels of
Caitlyn L Holmes et al.
PloS one, 14(5), e0217221-e0217221 (2019-05-29)
The antibody response to pertussis vaccination in rheumatoid arthritis is unknown, a concerning omission given the relatively low efficacy of the pertussis vaccine, a rise in pertussis infections, and a general increased susceptibility to infection in rheumatoid arthritis. Additionally, the
Timothy J Bowden et al.
Biology, 9(12) (2020-12-02)
Oysters and clams are important for food security and of commercial value worldwide. They are affected by anthropogenic changes and opportunistic pathogens and can be indicators of changes in ocean environments. Therefore, studies into biomarker discovery are of considerable value.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service