Skip to Content
Merck
All Photos(1)

Key Documents

77262

Sigma-Aldrich

Pentadecafluorooctanoic acid ammonium salt

≥98.0% (NT)

Synonym(s):

Ammonium pentadecafluorooctanoate, Perfluorocaprylic acid ammonium salt, Perfluorooctanoic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)6COONH4
CAS Number:
Molecular Weight:
431.10
Beilstein:
4286466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (NT)

form

powder

functional group

carboxylic acid
fluoro

SMILES string

N.OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8HF15O2.H3N/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23;/h(H,24,25);1H3

InChI key

YOALFLHFSFEMLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecafluorooctanoicacid ammonium salt (Ammonium pentadecafluorooctanoate) is commonly used in the polymerization of fluorinated monomers. It is also used as a surfactant during chemical synthesis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, interfacial property, and application of new hybrid anion surfactant containing fluorocarbon and hydrocarbon chains
Kang EK, et al.
Journal of Industrial and Engineering Chemistry, 67, 72-79 (2018)
Hydrocarboxylation of terminal alkenes in supercritical carbon dioxide using perfluorinated surfactants
Tortosa-Estorach C, et al.
Chemical Communications (Cambridge, England), 26, 2789-2791 (2006)
Amanda Ode et al.
Environmental science and pollution research international, 20(11), 7970-7978 (2013-02-26)
In recent years, some perfluorinated compounds (PFCs) have been identified as potentially hazardous substances which are harmful to the environment and human health. According to limited data, PFC levels in humans could be influenced by several determinants. However, the findings
A K Rosenmai et al.
Toxicology and applied pharmacology, 266(1), 132-142 (2012-11-13)
Polyfluoroalkyl phosphate surfactants (PAPS) are widely used in food contact materials (FCMs) of paper and board and have recently been detected in 57% of investigated materials. Human exposure occurs as PAPS have been measured in blood; however knowledge is lacking
Anne Vested et al.
Environmental health perspectives, 121(4), 453-458 (2013-01-31)
Perfluorinated alkyl acids (PFAAs), persistent chemicals with unique water-, dirt-, and oil-repellent properties, are suspected of having endocrine-disrupting activity. The PFAA compounds perfluorooctanoic acid (PFOA) and perfluorooctane sulfonic acid (PFOS) are found globally in humans; because they readily cross the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service