Skip to Content
Merck
All Photos(4)

Key Documents

164194

Sigma-Aldrich

Heptafluorobutyric acid

98%

Synonym(s):

Edman Reagent No. 3, HFBA, Perfluorobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2COOH
CAS Number:
Molecular Weight:
214.04
Beilstein:
1426882
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

7 (vs air)

Quality Level

vapor pressure

~10 mmHg ( 25 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

120 °C/755 mmHg (lit.)

density

1.645 g/mL at 25 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)

InChI key

YPJUNDFVDDCYIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perfluorobutyric acid is an effective additive for zinc electrodeposition[1].

Application

Perfluorobutyric acid was used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins[2]. It was used as ion pairing reagent in preparative and analytical reserved phase HPLC of underivatised peptides[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Naldi et al.
Journal of chromatography. A, 1129(1), 73-81 (2006-08-05)
The application of reversed-phase high-pressure liquid chromatography under gradient conditions and electrospray ion trap mass spectrometry (LC-ESI-MS) to the analysis of global modification levels of core histones is described. The optimised LC-ESI-MS method was applied for the first time to
D R Harding et al.
International journal of peptide and protein research, 18(2), 214-220 (1981-08-01)
This report describes the successful use of the ether soluble, ion pairing reagents perfluoropropoinic and perfluorobutyric acid in the preparative and analytical reserved phase HPLC of underivatised peptides. The preparative separation of a 1-g sample of Pyr-His-Gly, the proposed anorexigenic
Zinc electrowinning from acidic sulphate solutions Part IV: effects of perfluorocarboxylic acids.
Tripathy BC, et al.
Journal of Electroanalytical Chemistry, 565(1), 49-56 (2004)
Huilun Chen et al.
International journal of environmental research and public health, 17(4) (2020-02-23)
Perfluoroalkyl carboxylic acids (PFCAs) are some of the most significant pollutants in human serum, and are reported to be potentially toxic to humans. In this study, the binding mechanism of PFCAs with different carbon lengths to human serum albumin (HSA)
Kaberi P Das et al.
Toxicological sciences : an official journal of the Society of Toxicology, 105(1), 173-181 (2008-05-31)
Perfluorobutyrate (PFBA) is a perfluoroalkyl acid (PFAA) found in the environment. Previous studies have indicated developmental toxicity of PFAAs (perfluorooctane sulfonate [PFOS] and perfluorooctanoate [PFOA]); the current study examines that of PFBA. PFBA/NH4(+) was given to timed-pregnant CD-1 mice by

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service