All Photos(2)
About This Item
Linear Formula:
HSC6H4CO2H
CAS Number:
Molecular Weight:
154.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
form
solid
mp
215-224 °C (lit.)
SMILES string
OC(=O)c1ccc(S)cc1
InChI
1S/C7H6O2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H,(H,8,9)
InChI key
LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-mercaptobenzoic acid (4-MBA) is a probe molecule with thiol and carboxylic groups that form a self-assembled monolayers (SAMs) which can be used for the development of surface enhanced raman spectroscopy (SERS) sensors.
Application
Gold nanostructures functionalized with 4-Mercaptobenzoic acid (4-MBA) may exhibit improve catalytic activity. Several studies report 4-MBA as a model analyte for surface enhancement Raman scattering (SERS) substrates.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Surface enhanced Raman scattering of 4-mercaptobenzoic and haemoglobin adsorbed on self assembled Ag Monolayer films with different shapes
Zhu S, et al.
Applied Physics. A, Materials Science & Processing, 1075-1083 (2014)
Damiano Genovese et al.
Frontiers in chemistry, 8, 71-71 (2020-03-17)
The detection of the Cerenkov radiation (CR) is an emerging preclinical imaging technique which allows monitoring the in vivo distribution of radionuclides. Among its possible advantages, the most interesting is the simplicity and cost of the required instrumentation compared, e.g.
Electrochemical determination of bisphenol A with a glassy carbon electrode modified with gold nanodendrites
Chen WY, et al.
Microchimica Acta, 182, 703-709 (2015)
Synthesis and characterization of a material derived from 4-mercaptobenzoic acid: A novel platform for oligonucleotide immobilization.
Alves R, et al.
Talanta, 165, 69-75 (2017)
Journal of Molecular Structure, 976(1-3), 297-300 null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service