Skip to Content
Merck
All Photos(1)

Documents

755729

Sigma-Aldrich

(±)-Nicotine-(pyridine-d4)

≥98 atom % D, ≥98% (CP)

Synonym(s):

2,3,4,6-Tetradeuterio-5-(1-methylpyrrolidin-2-yl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D4H10N2
CAS Number:
Molecular Weight:
166.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

density

1.045 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1nc([2H])c(c([2H])c1[2H])C2CCCN2C

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/i2D,4D,6D,8D

InChI key

SNICXCGAKADSCV-MNYIHESISA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service