Skip to Content
Merck
All Photos(1)

Key Documents

755680

Sigma-Aldrich

Strontium aluminate

-100 mesh, 99.5% trace metals basis

Synonym(s):

Aluminum strontium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrAl2O4
CAS Number:
Molecular Weight:
205.58
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.5% trace metals basis

form

powder

particle size

-100 mesh

SMILES string

O=[Al]O[Sr]O[Al]=O

InChI

1S/2Al.4O.Sr

InChI key

YDLZPNUJQDLFBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazin; P. E.; Poltavets; V. V.; Tretyakov; Y. D.; Jansen; M.; Freitag; B.; Mader; W.
Physica C, 280, 253-253 (1997)
Toufic El Arnaout et al.
Scientific reports, 7(1), 6384-6384 (2017-07-27)
Scanning electron microscopy and X-ray microtomography are useful methods for high resolution shape imaging. Visible microscopy is also common, however, developing a low-cost and customizable system for surface and shape investigation of optically active particles is challenging. In this work
Katsumata; T.; Sasajima; K.; Nabae; T.; Komuro; S.; Morikawa; T.
Journal of the American Ceramic Society. American Ceramic Society, 81, 413-413 (1998)
Matsuzawa; T.; Aoki; Y.; Takeuchi; N.; Murayama; Y.
Journal of the Electrochemical Society, 143, 2670-2670 (1996)
Zhou; L.; Jiao; H.; He; D.
Materials Chemistry and Physics, 127, 227-227 (2011)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service