Skip to Content
Merck
All Photos(3)

Key Documents

757357

Sigma-Aldrich

Lanthanum gallate, strontium and magnesium doped

greener alternative

powder, 0.3-0.6 μm, 99% trace rare earth metals basis

Synonym(s):

Gallium lanthanum magnesium strontium oxide, LSGM, LSGM 8282, Lanthanum Strontium Gallate Magnesite, Strontium and Magnesium doped Lanthanum Gallate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La0.8Sr0.2Ga0.8Mg0.2O3
CAS Number:
Molecular Weight:
237.29
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

Assay

99% trace rare earth metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

4-8 m2/g

particle size

0.3-0.6 μm
0.4-0.8 μm (d50)

mp

>400 °C

greener alternative category

General description

LSGM is an electrolyte material for intermediate temperature (below 800oC ) solid oxide fuel cells (IT-SOFC). LaGaO3 based pervoskite; doped with strontium and magnesium (LSGM) is known to exhibit high oxide ionic conductivity, on account of the high concentration and mobility of oxygen vacancies LSGM-8282 can be synthesized by solid state reaction between La2O3, MgO, SrCo3 and Ga2O at temperatures > 1000oC. Other known methods of producing LSGM are combustion synthesis, coprecipitation route, Pechini method for powder. Pechini method is popularly used in producing LSGM for solid oxide fuel cells (SOFCs).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

World-wide a number of activities are concerned with the optimization and development of cell materials and microstructures with the aim of reducing the solid oxide fuel cell (SOFC) operating temperatures. Reducing operating temperatures not only increases the life time of SOFC but also reduces cost of the total system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Preparation of Pure and Strontium-and/or Magnesium-Doped Lanthanum Gallate Powders
Tas A, et al.
Journal of the American Ceramic Society. American Ceramic Society, 83(12), 2954-2960 (2000)
Huang, P-N.; et al.
Journal of the Electrochemical Society, 143, 1644-1644 (1996)
SOFC composite electrolyte based on LSGM-8282 and zirconia or doped zirconia from zircon concentrate
Rahmawati F, et al.
International Journal of Minerals, Metallurgy, and Materials, 19(9), 863-871 (2012)
Hwang, C-S.; et al.
Journal of Power Sources, 196, 1932-1932 (2011)
Oxygen surface exchange and diffusion in LaGaO 3 based perovskite type oxides.
Ishihara T, et al.
Solid State Ionics, 113, 593-600 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service