Skip to Content
Merck
All Photos(1)

Key Documents

749222

Sigma-Aldrich

Polyacrylamide

average Mn 150,000

Synonym(s):

PAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H5NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

Quality Level

mol wt

average Mn 150,000

mp

>200 °C

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • The influence of swelling on elastic properties of polyacrylamide hydrogels: Studies how swelling influences the mechanical properties of polyacrylamide hydrogels, relevant for biomaterials research and development (R Subramani et al., 2020).
  • Basics and recent advances of two dimensional-polyacrylamide gel electrophoresis: Reviews advancements in gel-based proteomics using polyacrylamide gel electrophoresis, essential for protein separation techniques in biochemical research (S Magdeldin et al., 2014).
  • Polyacrylamide gel electrophoresis: Provides comprehensive guidelines on using polyacrylamide gel electrophoresis for molecular biology applications, including its role in genotyping and molecular diagnostics (MR Green, J Sambrook, 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meital Bloch et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 47(5), 904-913 (2012-10-02)
Real time detection of biomarkers at the mucosal level is imperative for the prevention and efficient treatment of colorectal cancer. Cationized polyacrylamide (CPAA) with increasing charge densities was prepared by radical polymerization of acrylamide and different mol% ratios of N-acryloyl
Amandine Cuenca et al.
Physical review letters, 110(10), 108304-108304 (2013-03-26)
Pressure-driven flows of high molecular weight polyacrylamide solutions are examined in nanoslits using fluorescence photobleaching. The effective viscosity of polymer solutions decreases when the channel height decreases below the micron scale. In addition, the apparent slippage of the solutions is
Dammika P Manage et al.
Electrophoresis, 33(21), 3213-3221 (2012-10-03)
Electrophoresis is a powerful method that has seen a wide range of applications, often in automated genetic diagnostic instruments that require the use of a replaceable sieving matrix. The power and simplicity of electrophoresis as an analysis technique would be
Hyuck Joon Kwon et al.
Journal of the mechanical behavior of biomedical materials, 17, 337-346 (2012-11-07)
Many studies have demonstrated that sulfur-containing acidic groups induce chondrogenesis in vitro and in vivo. Recently, it is increasingly clear that mechanical properties of cell substrates largely influence cell differentiation. Thus, the present study investigated how mechanical properties of sulfonate-coated
Honghao Sun et al.
Bioconjugate chemistry, 23(11), 2247-2255 (2012-11-02)
Our ability to design receptor-targeted nanocarriers aimed at drug release after endocytosis is limited by the current knowledge of intracellular nanoparticle (NP) trafficking. It is not clear if NP size, surface chemistry, and/or targeting of cell surface receptors changes the

Articles

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service