Skip to Content
Merck
All Photos(2)

Key Documents

535311

Sigma-Aldrich

Poly(N-isopropylacrylamide)

Mn ~40,000

Synonym(s):

NIPAM polymer, PNIPAM, Polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H11NO)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

Mn 20,000-60,000
Mn ~40,000

mp

96 °C

SMILES string

CC(C)NC(=O)C=C

InChI

1S/C6H11NO/c1-3-5-7-6(8)4-2/h4H,2-3,5H2,1H3,(H,7,8)

InChI key

WDFKEEALECCKTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thermosensitive polymer, can be used to form a hydrogel. Aqueous polymer solution undergoes a phase transition from a soluble to an insoluble state when the temperature is raised to ca. 32 °C.

Physical form

Thermosensitive polymer, hydrogel

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Xia et al.
Chemical communications (Cambridge, England), 49(25), 2566-2568 (2013-02-21)
Site-specific protein conjugates with RAFT polymers were synthesized using expressed protein ligation. Stable micelles were formed from both linear block copolymer and Y-shaped conjugates.
Kornelia Gawlitza et al.
Physical chemistry chemical physics : PCCP, 14(27), 9594-9600 (2012-06-12)
The aim of the present work is the use of a water soluble enzyme in an organic solvent, still with a pronounced catalytic activity. Therefore, lipase B from Candida antarctica (CalB) is immobilized within micron-sized thermosensitive p-NIPAM hydrogel particles using
Jinseok Byun et al.
Chemical communications (Cambridge, England), 48(74), 9278-9280 (2012-08-09)
We report a novel approach for the fabrication of a photo-responsive surface with fast and reversibly switchable wettability between hydrophobicity and hydrophilicity induced by a photothermal effect.
Pengju Pan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(40), 14347-14356 (2012-09-28)
Linear and miktoarm star-shaped diblock copolymers consisting of single-stranded DNA and poly(N-isopropylacrylamide) (PNIPAAm) with various compositions were synthesized via atom transfer radical polymerization and click chemistry. The temperature-responsive phase transition behavior, micellization, was systematically examined using UV-vis spectrometry, high-sensitivity differential
Albert J de Graaf et al.
Journal of controlled release : official journal of the Controlled Release Society, 162(3), 582-590 (2012-09-14)
In this paper it is shown that when a thermosensitive hydrogel based on poly(N-isopropylacrylamide)-poly(ethylene glycol)-poly(N-isopropylacrylamide) (pNIPAm-PEG-pNIPAm) was transferred into water, flower-like micelles were continuously released as long as the medium was regularly refreshed. On the other hand, if the medium

Articles

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service