Skip to Content
Merck
All Photos(1)

Key Documents

715352

Sigma-Aldrich

Phenyl vinyl-1,2-13C2 sulfide

99 atom % 13C

Synonym(s):

(Phenylthio)ethylene-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5S13CH=13CH2
CAS Number:
Molecular Weight:
138.20
MDL number:
UNSPSC Code:
12352005

isotopic purity

99 atom % 13C

form

liquid

bp

94-95 °C/25 mmHg

density

1.057 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

[13CH2]=[13CH]Sc1ccccc1

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Licensed from Highlands Stable Isotopes Corp.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service