Skip to Content
Merck
All Photos(2)

Key Documents

703788

Sigma-Aldrich

Sodium tert-butoxide

99.9%

Synonym(s):

Sodium 2-methylpropan-2-olate, Sodium t-butoxide, Sodium tert-butanolate, Sodium tert-butylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOC(CH3)3
CAS Number:
Molecular Weight:
96.10
Beilstein:
3654215
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9%

form

powder

density

1.025  g/cm3 at 68 °F

SMILES string

[Na+].CC(C)(C)[O-]
[Na+].CC(C)(C)[O-]

InChI

1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1
1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

MFRIHAYPQRLWNB-UHFFFAOYSA-N
MFRIHAYPQRLWNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium tert-butoxide can be used as:
  • A promoter for the synthesis of biaryls by C-H bond arylation of aromatic compounds with haloarenes.
  • A base in palladium catalyzed amination reactions.
  • A base in the synthesis of aryl tert-butyl ethers from unactivated aryl halides in presence of palladium catalyst.
  • In the desulfonylation of N-indoles and N-azaindoles.
  • As a base in the synthesis of sodium acrylates from olefins and CO2.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of acrylates from olefins and CO2 using sodium alkoxides as bases
Manzini S, et al.
Catalysis Today, 281(10), 379-386 (2017)
Desulfonylation of Indoles and 7-Azaindoles Using Sodium tert-Butoxide
Chaulet C, et al.
Synlett, 2010(10), 1481-1484 (2010)
tert-Butoxide-Mediated C-H Bond Arylation of Aromatic Compounds with Haloarenes
Yanagisawa S and Itami K
ChemCatChem, 3(5), 827-829 (2011)
β-Hydrogen-Containing Sodium Alkoxides as Suitable Bases in Palladium-Catalyzed Aminations of Aryl Halides
Prashad M, et al.
The Journal of Organic Chemistry, 65(8), 2612-2614 (2000)
Palladium-catalyzed formation of aryl tert-butyl ethers from unactivated aryl halides
Parrish CA and Buchwald SL
The Journal of Organic Chemistry, 66(7), 2498-2500 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service