Skip to Content
Merck
All Photos(2)

Documents

526460

Sigma-Aldrich

Xantphos

97%

Synonym(s):

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H32OP2
CAS Number:
Molecular Weight:
578.62
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

reagent type: ligand
reaction type: Hydroformylations

reagent type: ligand
reaction type: Miyaura Borylation Reaction

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

224-228 °C (lit.)

functional group

phosphine

SMILES string

CC1(c2c(Oc3c1cccc3P(c4ccccc4)c5ccccc5)c(P(c6ccccc6)c7ccccc7)ccc2)C

InChI

1S/C39H32OP2/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32/h3-28H,1-2H3

InChI key

CXNIUSPIQKWYAI-UHFFFAOYSA-N

General description

Xantphos is a bidentate ligand that is employed in various metal-catalyzed reactions, including cross-couplings, Buchwald-Hartwig cross-coupling reactions, C-X bond formation, hydroformylation, Miyaura borylation reaction, Stille coupling, Suzuki-Miyaura coupling, (hetero)arylation, amination, and amidation.

For small scale and high throughput uses, product is also available as ChemBeads (928356)

Application

Ligand used in a synthesis of heterocycles by palladium-catalyzed C-N cross coupling of 3-bromothiophenes with 2-aminopyridines. Also used in a ruthenium-catalyzed alkylation of active methylene compounds with alcohols.
Metal chelating ligand for catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Manley et al.
Organic letters, 6(14), 2433-2435 (2004-07-02)
[reaction: see text] An alternative to the Friedlander condensation for the synthesis of naphthyridinones and quinolinones has been discovered. Palladium-catalyzed amidation of halo aromatics substituted in the ortho position by a carbonyl functional group or its equivalent with primary or
Sarah Keller et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 17(4), 375-385 (2018-02-20)
The heteroleptic complexes [Cu(tBu-xantphos)(bpy)][PF6] and [Ag(tBu-xantphos)(bpy)][PF6], where tBu-xantphos = 9,9-dimethyl-4,5-bis(di-tert-butylphosphino)xanthene and bpy = 2,2'-bipyridine have been synthesized and their photophysical properties investigated. Single crystal X-ray diffraction studies of the compounds under ambient and increased pressure are presented; increase in pressure
Ligand Bite Angle Effects in Metal-catalyzed C-C Bond Formation.
P W van Leeuwen et al.
Chemical reviews, 100(8), 2741-2770 (2001-12-26)
Masoud Sadeghzadeh et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-20)
Monocarboxylate transporters 1-4 (MCT1-4) are involved in several metabolism-related diseases, especially cancer, providing the chance to be considered as relevant targets for diagnosis and therapy. [18F]FACH was recently developed and showed very promising preclinical results as a potential positron emission
Synthesis, 2794-2794 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service