Recommended Products
form
powder
Quality Level
concentration
~13% Mg
loss
≤10% loss on drying
SMILES string
O.[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O
InChI
1S/2Mg.2HO3Si.O3Si.H2O/c;;3*1-4(2)3;/h;;2*1H;;1H2/q2*+2;2*-1;-2;
InChI key
TUKQLEWOUPCTOS-UHFFFAOYSA-N
General description
Sepiolite is a hydrated magnesium silicate that belongs to the class of sepiolite-palygorskite. Its properties include high absorption capacity and good rheological behavior.
Application
Sepiolites can be used as absorbents for potential applications in catalytic systems and wastewater treatment.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thermal and morphological characterisation of organically modified sepiolite
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 107(1-2), 161-168 (2008)
Adsorption kinetics and thermodynamics of an anionic dye onto sepiolite
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 101(3), 388-396 (2007)
Experimental and Atomic Modeling of the Adsorption of Acid Azo Dye 57 to Sepiolite
Clays and Clay Minerals, 66(5), 426-437 (2018)
Journal of hazardous materials, 217-218, 11-18 (2012-04-03)
Cuprous oxide is firstly created on acidized sepiolite (AS) by a simple deposition method for photocatalytic degradation of the red water produced from 2,4,6-trinitrotoluene (TNT) manufacturing. X-ray diffraction (XRD), field-emission scanning electron microscopy (FE-SEM), ultraviolet-visible diffuse reflection absorptive spectroscopy (UV-vis/DRS)
Journal of nanoscience and nanotechnology, 11(1), 382-385 (2011-03-31)
Sepiocite, a synthetic sepiolite-like nanoclay, was derived from hydrotalcite-like Mg2Al(CO3)0.5-layered double hydroxide (LDH) under phase transformation at 270 +/- 3 degrees C. The crystal structure of sepiocite is conceptually very similar to that of sepiolite derived from montmorillonite clay because
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service